Plasmid DNA Purification
PureLink™ HiPure Plasmid Filter Midi/Maxi Starter Kit (2 reactions of midi and maxi)
Transfection Grade Large-Scale Plasmid Midi/Maxi Preps (>350 ug, 0.1 EU/ug). 
Plasmid purification with higher yields, more effective lysate filtration, and higher final concentrations to consistently produce transfection-grade DNA for any application.
ChargeSwitch®-Pro Miniprep Kit (10 preps)
Transfection Grade Small-Scale Plasmid Miniprep (<50ug, 0.1 EU/ug). 
Purify Transfection grade DNA in only 30 minutes.  Simple Bind, Wash, Elute protocol without the use of inhibitory reagents such as ethanol, chaotropic salts, or organic solvents.
ChargeSwitch® Pro Filter  Plasmid Mini Kit (10 preps)
ChargeSwitch® Pro Filter  Plasmid Midi/Maxi Kit (2 reactions of midi and maxi) 
NEW-Transfection Grade Plasmid Kits Mini, Midi, Maxi (0.01-0.1 EU/ug)
Our innovative redesign of the CST Pro kits offers the best of both brands with the lysate filtering capability of our PureLink™ kits with the speed of ChargeSwitch®.
Get Transfection Grade plasmid in as little as 15 minutes and our new ER reduction buffer.  The simple Bind, Wash, Elute protocol is compatible with centrifugation or vacuum filtration.  Use with our new EveryPrep™ Universal manifold and elute directly into your tubes using a vacuum.
RNA Purification
PureLink™ RNA Mini Kit (10 preps)
Rapid purification of total RNA from animal, plant, yeast cells, bacteria, and blood, combining a safe guanidine-isothiocyanate lysis with the speed, purity, and ease of use of silica-membrane purification. Purified RNA in less than 20 minutes. Highest binding capacity, up to 1 mg of RNA.
Genomic DNA Purification
PureLink™ Genomic DNA Mini Kit (10 preps)
High-yield, high-purity DNA extractions from blood, tissues, cells, bacteria, swabs, and blood spots using an optimized spin-column membrane and buffer formulations.