Search Thermo Fisher Scientific
Site | Enzyme |
---|---|
2161 | AccI |
4506 | AflIII |
2690 | AgeI |
2237 | BamHI |
943 | BbvCI |
3755 | BfrBI |
2853 | BlpI |
3160 | BsiWI |
3011 | BsmI |
3089 | BspMI |
2017 | BsrGI |
2570 | EcoRI |
2162 | HincII |
2175 | HindIII |
3757 | NsiI |
4606 | PciI |
2535 | PmlI |
3220 | RsrII |
3318 | SacII |
2160 | SalI |
4490 | SapI |
556 | SgrAI |
953 | XbaI |
2154 | XhoI |
Enzyme |
---|
AarI |
AleI |
ApaI |
AsiSI |
AvrII |
BbsI |
BclI |
BglII |
BplI |
BplI' |
BsaBI |
BstBI |
BstXI |
BstZ17I |
FseI |
FspAI |
HpaI |
MfeI |
MluI |
NcoI |
NotI |
NruI |
PacI |
PflMI |
PmeI |
PshAI |
PsiI |
PspOMI |
PsrI |
PsrI' |
SanDI |
SbfI |
SfiI |
SnaBI |
SrfI |
SwaI |
TaqII' |
XcmI |