Separation and purification of the target DNA sequence prior to cloning is critical to obtaining the desired recombinant clones. We offer a wide range of reagents for DNA electrophoresis to separate and excise your band of interest from hassle-free E-Gel CloneWell precast gel to low melting UltraPure agarose. Choosing the right reagents will allow you to achieve optimal results in your experiment.
Which DNA Gel is Right for You?
Low melting point agarose for pour your own gels | Convenient & sensitive precast gel | Precast gel designed for easy band capture ready for cloning | |
---|---|---|---|
Top Seller | ![]() | ||
Prep Time | 30 min | Ready to use | Ready to use |
Run Time | 60 min | 10 min | 15 min |
Sample Purification Time For Cloning* | 30 min | 30 min | Ready to use |
Gel Dye | Add your own dye | Proprietary Stain | SYBR Safe |
Sensitivity | Variable | Ultra Sensitive (> 1 ng) | Highly Sensitive (>5 ng) |
# of Lanes | Variable | 11 | 12 |
% Gels Available | Variable | 1%, 2% & 4% | 0.8% |
Nonhazardous / Environmentally Friendly Options | ![]() | ![]() | |
Pack Sizes Available | 50 & 100 g | 10 & 20 gels | 18 gels |
Order Now | Order Now | Order Now |
*The amount of time required to excise the DNA band from the gel and purify the DNA from the agarose prior to use for cloning. Learn more.
For Research Use Only. Not for use in diagnostic procedures.