Looking for best choice in agarose gels to analyze PCR products, restriction digests, or plasmid preparation? Look no further. We offer you a wide selection of reagents for your routine electrophoresis, ranging from precast agarose gels with fast and consistent separation of DNA in the 20 bp–10 kb size range, to casting your own gels with UltraPure agarose.
Which DNA gel is right for you?
Order info | Order info | Order info | |
Pour-your-own gel reagents | Broadest range of precast bufferless gels | Ultimate speed & sensitivity | |
Learn more › | Learn more › | Learn more › | |
Top seller | |||
Prep time | 30 min | Ready to use | Ready to use |
Run time | 60 min | 15–30 min | 10 min |
Sensitivity | Variable | Highly Sensitive ( >5 ng) | Ultra Sensitive ( >1 ng) |
# of lanes | Variable | 12–18 | 11 |
% gels available | Variable | 0.8%, 1.2%, 2% & 4% | 1%, 2% & 4% |
Nonhazardous/ environmentally friendly options | |||
Pack sizes available | 100 g, 500 g | 18 gels | 10 gels, 20 gels |
For Research Use Only. Not for use in diagnostic procedures.