TrackIt™ DNA Ladders
Convenient, ready-to-use DNA ladders updated to deliver clear bands for DNA sizing and approximate quantification.
  • Sharp, clear bands – chromatography purified fragments for consistent and reliable results and stable at room temperature
  • Versatile—broad size range from 10 bp to 12 Kb
  • Ready-to-use—premixed in loading buffer and stable at room temperature
  • Convenient—supplied with loading dye for sample DNA
Request a free sample
Select your TrackIt DNA Ladder
TrackIt DNA Ladders are ready to use and available in popular size ranges. TrackIt DNA Ladders all have reference bands (indicated in bold in the image below) for easy orientation. Select the ladder of interest in Table 1 below to go to ordering page.
Table 1. TrackIt DNA Ladder selection guide
Ordering information
Resources
Nucleic acid education
Learn about gel electrophoresis basics, workflows, considerations, applications, and troubleshooting in separation and analysis of nucleic acids.
Related products
Support
Nucleic Acid Purification and Analysis Support Center
Find tips, troubleshooting help, and resources for your nucleic acid purification and analysis applications.
Product support documentation
Search for manuals, protocols, Material Safety Data Sheets, product literature and certificates by catalog number or product name.
Contact us
Email or call our Technical Application Scientists for additional questions regarding DNA ladders