E-Gel 48 and 96-well Agarose Gels
E-Gel 48 and 96 well gels are ideal for high-throughput analysis of PCR products, plasmid preparations, and restriction digests. E-Gel 48 gels include two rows each containing 24 samples and 2 molecular weight marker wells with a 3.2 cm length. E-Gel 96 gels contain 96 sample lanes and 8 marker lanes, staggered to provide a 1.6-cm run length with resolution between 100 bp and 10 kb. This staggered-well format is compatible with most robotic loading systems.
Ordering information
Product Stains #Sample wells* Run time (min) Resolution Qty (gels) Cat. No.
E-Gel 48 Agarose Gels, 1% SYBR Safe 48 sample,
4 marker**
20 400 bp–10 kb 8 G820801
E-Gel 48 Agarose Gels, 1% SYBR Safe 48 sample,
4 marker**
20 400 bp–10 kb 4 x 8 G820841
E-Gel 48 Agarose Gels, 2% SYBR Safe 48 sample,
4 marker**
20 50 bp–3 kb 8 G820802
E-Gel 48 Agarose Gels, 2% SYBR Safe 48 sample,
4 marker**
20 50 bp–3 kb 4 x 8 G820842
E-Gel 96 Gels with SYBR Safe DNA Gel Stain, 1% SYBR Safe 96 sample,
8 marker**
12 1 bp–10 kb 8 G720801
E-Gel 96 Gels with SYBR Safe DNA Gel Stain, 1% SYBR Safe 96 sample,
8 marker**
12 1 bp–10 kb 4 x 8 G720841
E-Gel 96 Gels with SYBR Safe DNA Gel Stain, 2% SYBR Safe 96 sample,
8 marker**
12 100 bp–2 kb 8 G720802
E-Gel 96 Gels with SYBR Safe DNA Gel Stain, 2% SYBR Safe 96 sample,
8 marker**
12 100 bp–2 kb 4 x 8 G720842
E-Gel 48 Agarose Gels, 1% Ethidium bromide 48 sample,
4 marker**
20 400 bp–10 kb 8 G800801
E-Gel 48 Agarose Gels, 2% Ethidium bromide 48 sample,
4 marker**
20 50 bp–3 kb 8 G800802
E-Gel 48 Agarose Gels, 4% Ethidium bromide 48 sample,
4 marker**
20 10 bp–400 bp 8 G800804
E-Gel 96 Agarose Gels, 1% Ethidium bromide 96 sample,
8 marker**
12 1 kb–10 kb 8 G700801
E-Gel 96 Agarose Gels, 2% Ethidium bromide 96 sample,
8 marker**
12 100 bp–2 kb 8 G700802
* Sample volume is 20 µL except on E-Gel 48 Gels, which have 15 µL sample volume.
** Wells compatible with multichannel pipettor loading.