High-throughput DNA electrophoresis and documentation system
The E-Gel Power Snap Plus Electrophoresis System comprises two integrated modules:
- Gel electrophoresis device for rapid nucleic acid analysis and real-time view using blue light transilluminator. Compatible with 11-, 22-, 48-, and 96-well E-Gel precast agarose gels
- High-resolution camera instrument for experiment documentation, analysis, and export to Thermo Fisher Connect
High-throughput gel electrophoresis applications
Its high capacity and fast running time make the E-Gel Power Snap Plus System ideal for:
- High-throughput genotyping
- Cell population screening for quality control or mutations
- High-throughput clone library analysis and clone identification
- Any time high-throughput gel electrophoresis can help improve your workflow
Invitrogen E-Gel Power Snap Plus videos
Invitrogen E-Gel Power Snap Plus System features
Discover the features of this high-throughput DNA electrophoresis system including 96-well gel electrophoresis device and integrated gel imaging camera. When used with E-Gel precast agarose gels with SYBR Safe DNA Stain, the E-Gel Power Snap Plus System is a safer and greener option compared to conventional DNA electrophoresis.
E-Gel Power Snap Plus System
Learn how to perform high throughput gel electrophoresis, monitor DNA migration, and capture gel images with the E-Gel Power Snap Plus System. Find out about the image editing and image storage options.
iBright image analysis software
iBright Image Analysis Software analyzes data from E-Gel images captured by the integrated camera. The gel images and the results of data analysis can be stored on Thermo Fisher’s Connect Platform. iBright Image Analysis is included in the E-Gel Power Snap Plus Electrophoresis System.
E-Gel Power Snap Plus Electrophoresis System options
Molecular biology videos
Watch educational and insightful videos on molecular biology techniques.
Molecular biology webinars
View live and recorded webinars on research trends in molecular biology.
Molecular biology literature
Access white papers, application notes, and technical notes on molecular biology.
Precast gel and ladder selection tool
Find the right E-Gel precast gels and DNA ladders for your applications.
For Research Use Only. Not for use in diagnostic procedures.