RNA extraction
Whether you’re starting with cultured cells or or tissue samples, or working with plants, bacteria, or mammalian cells, Ambion RNA extraction reagents and kits offer the right product for your research. The Ambion brand includes legacy Invitrogen products such as trusted TRIzol Reagents and PureLink purification kits, alongside premier Ambion products such as Cells-to-CT, MagMAX, and RNAlater kits and reagents.
Which RNA extraction kit from blood sample is right for you?
  Order now Order now Order now Order now Order now Order now
  Optimized for liquid samples Specialized kit deplete non-nucleated erythrocytes Filter plate format with 96 columns, for HTP needs HTP optimized for purification from stabilized blood samples HTP format,capable of recovering RNA of all sizes (including microRNAs) High RNA yield directly from whole blood, no need to isolate WBCs
  TRIzol LS PureLink Total RNA Blood Kit PureLink Pro 96 total RNA Purification Kit MagMAX for Stabilized Blood Tubes RNA Isolation Kit MagMAX mirVana Total RNA Isolation RiboPure Blood Kit
Top Seller          
Compatible stabilization reagents EDTA, Heparin, Citrate, RNAlater EDTA, Heparin, Citrate EDTA, Heparin, Citrate Tempus or PaxGene tubes EDTA or Citrate EDTA, Heparin, Citrate, RNAlater
Stabilization reagent included          
RNAlater
Isolation method High purity organic extraction (requires alcohol precipitation) Fast, convenient silica column Fast, convenient silica columns in a plate format Scalable, flexible format with magnetic beads Scalable, flexible format with magnetic beads Highest purity & convenience; includes both organic extraction & silica column (no precipitation required)
Prep time 1 hour 20 minutes   12 tubes in 2 hours 96 samples in <1 hr 30 minutes
High throughput compatible      
For more info on working with blood please review our technical articles:
Supplemental Protocols
TechNotes Articles
Purifying RNA from cultured cells has traditionally been a highly repetitive, time- and labor-intensive procedure, often involving the use of toxic organic chemicals. The products developed by our RNA experts allow you to isolate RNA of maximum yield, purity, and integrity using several technology formats (organic extraction, glass-fiber columns, magnetic beads, and combinations of these).
You can also analyze cultured cell lysates without first isolating the RNA. In significantly less time, you’ll get qPCR results equivalent to - or in some cases better than - those from purified RNA. The Invitrogen products such as  Cells-to-CT and Single Cell-to-CT product lines offer tremendous performance and handling advantages.
Which RNA Isolation Kit from Cultured Cells is Right for You?
  Order now Order now Order now Order now Order now Order now
  Highly pure, intact RNA Fast and convenient column based format for RNA (>200 nt) Fast and convenient column based, filter plateformat for 96 samples (Rna > 200 nt) Fast and convenient column based method from limited sample input (Rna >200 nt) Fast and convenient method for small RNA species Complete system for cells to qRT-PCR without RNA purification
  TRIzol Reagent PureLink RNA Mini Kit PureLink Pro 96 total RNA Purification Kit PureLink RNA Micro Scale Kit MirVana miRNA Isolation Kit Cells-to-CT kits
Top seller    
Prep time 1 hour 20 minutes 20 minutes 20 minutes 30 minutes <10 minutes
Cell lysis method High purity organic extraction (requires alcohol precipi-tation) Nontoxic guanidine-isothio-cyanate lysis Nontoxic guanidine-isothio-cyanate lysis Nontoxic guanidine-isothio-cyanate lysis Organic extraction followed by alcohol precipitation Cells-to-CT lysis
Amount of Starting material Up to 1x107 cells 5x106 to 1x108 cells 5x106 to 1x108 cells Up to 5x105 cells 102-107 cultured cells 10-100,000 cells
Appli-cation All RNA expression methods qRT-PCR, NGS qRT-PCR, NGS qRT-PCR, NGS Micro RNA analysis qRT-PCR
Preserve and Stabilize Your RNA
RNAlater Solution is widely used for the preservation of RNA in intact animal tissues, organs, and cells, and bacteria. It eliminates the need to immediately process samples for RNA isolation or to freeze them in liquid nitrogen for later processing.
Technical Resources
Which RNA Extraction Kit from FFPE Samples is Right for You?
  Order now Order now
  Low-throughput isolation of RNA and DNA from the same FFPE section Scalable isolation of RNA and DNA from the same FFPE section
  RecoverAll Total Nucleic Acid Isolation Kit for FFPE MagMAX FFPE DNA/RNA Ultra Kit
Prep Time 4.5 hours (total time for 16 samples)
2.5 hrs hands-on time
4 hours (total time for 96 preps)
1 hr hands-on time
Final product extracted total RNA, microRNA, gDNA total RNA, microRNA, gDNA
Isolation method column purification mag bead purification
Deparaffinization required
Automation compatible (post deparffinization)  
(Scripts for KF Flex and Duo Prime)
Compatible FFPE sample range up to 4 x 20 micron sections
(300 mm2 tissue)
up to 2 x 20 micron sections
(300 mm2 tissue)
Sample volume ~50µL purified DNA
~50µL purified RNA
~50uL purified DNA
~50µL purified RNA
Compatible with Ion AmpliSeq DNA panels (e.g. Cancer Hotspot Panel v2)
Compatible with Ion AmpliSeq RNA panels (e.g. RNA cancer and RNA fusion)
Why is RNA Isolation From FFPE such a Challenge?
The recovery of quality RNA from FFPE specimens can be quite challenging. The fixation process causes cross-linkage between nucleic acids and proteins, and covalently modifies RNA by the addition of monomethyl groups to the bases. As a result, the molecules are rigid and susceptible to mechanical shearing, and the cross-links may compromise the use of RNA as a substrate for reverse transcription. Therefore, in order to utilize FFPE tissues as a source for gene expression analysis, a reliable method is required for extraction of RNA from the cross-linked matrix.
How can you Increase your Chances of Success With FFPE?
There are a number of factors that can impact the overall quality and yield of RNA isolated from FFPE tissues. Here are recommendations to address several key factors:
  1. Upstream tissue procurement and tissue specimen preparation—if possible, tissues should be fixed within one hour of surgical resection. Extensive degradation of RNA can occur before completion of the fixation process. The optimal fixation time is 12–24 hours, using neutral-buffered formalin or paraformaldehyde. Fixed tissues should be thoroughly dehydrated prior to the embedding process.
  2. Block storage—storage of blocks without cut faces, when possible, prevents ongoing damage from exposure to atmospheric oxygen, water, and other environmental factors such as light and infestation (fungus, insects, etc.).
  3. Tissue type, size, and amount being used for RNA isolation—the recommended tissue thickness is 10–20 µm The number of sections used is determined by the tissue type (which impacts cell density) and surface area (recommended size: 50–300 mm2). Excess starting material can cause filter clogging, resulting in poor yield.
  4. Excessive amount of paraffin used for embedding tissues—when possible, excess paraffin should be trimmed away prior to starting the purification protocol. For xylene-based purification methods, two xylene treatments at room temperature should be sufficient for complete deparaffinization. If desired, a more rigorous 37–55°C treatment can be performed for up to 30 minutes. After the xylene deparaffinization, it is crucial that the 100% ethanol is completely removed and the pellets are dry after the two 100% ethanol washes. The magnetic bead method employs novel chemistries to deal with the paraffin that limits input to 20 µm sections.
The MagMAX FFPE DNA/RNA Ultra Kits are designed for rapid, efficient, and automatable isolation of total RNA and DNA from FFPE samples using Invitrogen MagMAX magnetic particles. Invitrogen MagMAX FFPE NA kits offer faster workflows and use less toxic reagents, without sacrificing quality in the process. These kits help deliver nucleic acid yield and purity comparable to the best-in-class RecoverAll filter-based system. The MagMAX FFPE NA kits outperform products with similar protocols, and achieve RNA and DNA yields that are comparable with longer, more hazardous, and lower-throughput protocols.
The RecoverAll Total Nucleic Acid Isolation Kit procedure requires about 45 minutes of hands-on time and can easily be completed in less than 1 day when isolating RNA. FFPE samples are deparaffinized using a series of xylene and ethanol washes. Next, they are subjected to a rigorous protease digestion with an incubation time tailored for recovery of either RNA or DNA. The nucleic acids are purified using a rapid glass-filter methodology that includes an on-filter nuclease treatment, and are eluted into either water or the low-salt buffer provided.
Technical Resources
TechNotes Articles
Tips from the Bench
Your Data
Application Note
Product Bulletin
Which Plant RNA Isolation Kit is Right for You?
  Order now Order now Order now Order now
  Great for difficult samples (conifer tissue & seeds) Quick & easy to use Efficient recovery of microRNA & small RNA Rapid & fully automated
  Plant RNA Reagent PureLink RNA Mini Kit mirVana miRNA Isolation Kit MagMAX-96 Total RNA Isolation Kit
Top seller      
RNA types isolated large RNA molecules only large RNA molecules only Small & large RNA molecules large RNA molecules only
Prep time 60 minutes <20 minutes 30 minutes <45 minutes
Amount of starting material Up to 1 g <250 mg 0.5 to 250 mg Up to 10 mg
Isolation method High purity organic extraction (requires alcohol precipitation) Fast, convenient silica column Highest purity & convenience; includes organic extraction & silica column (no precipitation required) Scalable, flexible format with magnetic beads
High throughput compatible      
Compatible products   Ambion Plant RNA Isolation Aid
Removes common plant contaminants
Ambion Plant RNA Isolation Aid
Removes common plant contaminants
Ambion Plant RNA Isolation Aid
Removes common plant contaminants
Remove Polysaccharides
One of the major issues with plant RNA isolation is the presence of problematic biomolecules, such as polyphenolic compounds and polysaccharides. The Ambion Plant RNA Isolation Aid minimizes many of these contaminants with the addition of polyvinylpyrrolidone, a high molecular weight polymer that binds to polyphenolics and polysaccharides and a pre-extraction spin step.
Preserve and Stabilize Your RNA
RNAlater Solution is widely for the preservation of RNA in intact animal tissues, organs, and cells, and bacteria. It minimizes the need to immediately process samples for RNA isolation or to freeze them in liquid nitrogen for later processing.
Technical Resources
Which tissue RNA extraction kit is right for you?
  Order now Order now Order now Order now Order now Order now Order now
  Gold standard for highly pure, intact RNA Simple, reliable, rapid method Fast and convenient column based, filter plateformat for 96 samples (Rna > 200 nt) Fast and convenient column based method from limited sample input (Rna >200 nt) Fast and convenient method for small RNA species Highest purity TRIzol + column purification, no precipitation required HTP format, capable of recovering RNA of all sizes (including microRNAs)
  TRIzol Reagent PureLink RNA Mini Kit PureLink Pro 96 total RNA Purification Kit PureLink RNA Micro Scale Kit mirVana miRNA Isolation Kit TRIzol Plus RNA Purification System MagMAX mirVana Total RNA Isolation
Top Seller            
Isolation method High purity organic extraction (requires alcohol precipitation) Fast, convenient silica column Fast Convenient silica column in a filter plate format Fast Convenient micro silica columns Organic extraction combined with convenient silica columns Highest purity and convenience; includes both organic extraction and silica column (no precipitation required) Scalable, flexible format with magnetic beads
Prep time 1 hour 20 minutes 20 minutes 20 minutes 30 minutes 1 hour <1 hour
Amount of starting material Up to 100 mg 10 mg to 100 mg 10 cells to 10 mg up to 10 mg up to 250 mg tissue Up to 100 mg Up to 50 mg
High throughput compatible          
Prep size 100 preps 50 preps 96 preps 50 preps 96 preps 50 preps 96 preps
RNA Yields From Animal Tissues
RNA content can vary widely between tissues, cell types, physiological states, etc. This chart provides general guidelines for estimating RNA yields from a variety of cells and tissues.
Table 1. Typical total RNA yields from 1 mg of various tissues.
Preserve and Stabilize Your RNA
RNAlater Solution is widely used for the preservation of RNA in intact animal tissues, organs, and cells, and bacteria. It eliminates the need to immediately process samples for RNA isolation or to freeze them in liquid nitrogen for later processing.
Technical Resources
Which Yeast RNA Extraction Kit is Right for You?
  Order now Order now Order now Order now
  Rapid & fully automated Quick & easy to use Designed for rigid yeast cell walls while maintaining expression profiles Fast and convenient column based, filter plateformat for 96 samples (Rna > 200 nt)
  Platinum Taq MagMAX for Microarrays Total RNA Isolation Kit PureLink RNA Mini Kit RiboPure-Yeast Kit PureLink Pro 96 total RNA Purification Kit
Top seller      
Includes specialized beads for cell lysis No No Zirconia beads No
Isolation method Scalable, flexible format for highest purity; includes organic extraction and magnetic beads Fast, convenient silica column Highest purity and convenience; includes both organic extraction and silica column Fast, convenient silica column in 96 well filter plate format
Prep time <1 hour 20 minutes 90 minutes (includes DNase treatment) 20 minutes
Amount of starting material Up to 5 x 106 cells Up to 5 x 108 cells Up to 3 x 108 cells 1.8 mL of fresh, log-phase yeast cells (OD660=0.6–0.8)
High throughput compatible      
Kit size 96 preps 50 preps 50 preps 96 preps
Technical Resources
Unlike eukaryotic cells, yeast and many bacterial strains are protected by cell walls that can be difficult to disrupt. As a first step, effective cell lysis (typically via mechanical disruption or enzymatic pretreatment) is required prior to any RNA isolation procedure.
A range of products developed by the Ambion RNA experts will help you isolate high yields of pure, intact RNA from bacteria and yeast.
Which Bacterial RNA Extraction Kit is Right for You?
  Order now Order now Order now Order now Order now
  No lysis procedures Fast & easy, 20 min Fast and convenient silica column based , filter plate format HTP, 45 min Ideal for hard-to-lyse gram + bacteria
  TRIzol Max Bacterial RNA Isolation Kit w/TRIzol & Max Bacterial Enhancement Reagent PureLink RNA Mini Kit PureLink Pro 96 total RNA Purification Kit MagMAX Total Nucleic Acid Isolation Kit RiboPure-Bacteria Kit
Top Seller        
Includes specialized beads for cell lysis No No No Zirconia beads Zirconia beads
Isolation method High purity organic extraction (requires alcohol precipitation) Fast, convenient silica column Fast, convenient silica column in 96 well filter plate format Scalable, flexible format with magnetic beads Highest purity & convenience; includes organic extraction & silica column (no precipitation required)
Prep time 1 hour 20 minutes 20 minutes 45 minutes 1 hour
Amount of starting material Up to 1 x 108 cells Up to 1 x 109 cells Up to 1 x 109 cells Up to 0.3 g solid samples or 175 µl liquid samples Up to 109 cells
High throughput compatible      
Kit size 100 preps 50 preps 96 preps 100 preps 50 preps
Technical Resources
Viral analysis of biological (animal, insect, plant, fungal, bacterial) and environmental (water, air, food) samples is increasingly conducted using molecular testing strategies that require viral RNA or DNA from the samples being tested. Purification of viral RNA poses unique challenges for getting good recovery and detection sensitivity. Ambion RNA experts have developed RNA purification products that are optimized to provide maximum viral RNA yield, purity, and integrity from a broad range of sample types in several format options.
"Highly concentrated viral RNA from the PureLink Viral RNA/DNA Mini Kit provides a tool for more sensitive and delicate downstream analysis on viral genomes”
Dongning Wang, PhD, in the laboratory of Feng Gao, MD, Duke Human Vaccine Institute, Duke University
Product Specifications
  Most Suitable Downstream Application
Product Format Real-Time RT-PCR Microarray Analysis Sequen-cing Northern Blot Cloning
Total RNA
PureLink Viral RNA⁄DNA Mini Kit Chaotropic salt-based lysis, silica filter      
RNAqueous Kit Chaotropic salt-based lysis, silica filter    
MagMAX AI⁄ND Viral RNA Isolation Kit Chaotropic salt-based lysis, magnetic particles      
MagMAX-96 AI⁄ND Viral RNA Isolation Kit Chaotropic salt-based lysis, magnetic particles, 96-well processing, automation compatible      
DNA/RNA
PureLink Pro 96 Viral RNA⁄DNA Purification Kit Chaotropic salt-based lysis, silica filter, 96-well processing, automation compatible        
MagMAX Total Nucleic Acid Isolation Kit Chaotropic salt-based lysis, magnetic particles, 96-well processing, automation compatible      
MagMAX-96 Viral RNA Isolation Kit Chaotropic salt-based lysis, magnetic particles, 96-well processing, automation compatible      
Dynabeads SILANE Viral NA Chaotropic salt-based lysis, magnetic beads, automation compatible          
iPrep PureLink Virus Kit Chaotropic salt-based lysis, magnetic particles, automation compatible        
MagMAX Viral RNA Isolation Kit Chaotropic salt-based lysis, magnetic particles        
MagMAX-96 Viral RNA Isolation Kit (5 x 96 rxns) Chaotropic salt-based lysis, magnetic particles, 96-well processing, automation compatible        
MicroRNA
mirVana miRNA Isolation Kit Chaotropic salt-based lysis, silica filter        
mirVana miRNA Isolation Kit (without phenol - must be purchased separately) Chaotropic salt-based lysis, silica filter        
TRIzol LS Reagent Lysis in TRIzol reagent, one-step reagent  
mRNA
Dynabeads mRNA DIRECT Kit Chaotropic salt-based lysis, magnetic beads      
Dynabeads mRNA DIRECT Micro Kit Chaotropic salt-based lysis, magnetic beads      
Resources
Nucleic Acid Purification and Analysis Support Center
Find tips, troubleshooting help, and resources for your nucleic acid purification & analysis applications.
DNA Purification Guide
RNA Isolation & Purification Guide
Which RNA extraction kit from blood sample is right for you?
  Order now Order now Order now Order now Order now Order now
  Optimized for liquid samples Specialized kit deplete non-nucleated erythrocytes Filter plate format with 96 columns, for HTP needs HTP optimized for purification from stabilized blood samples HTP format,capable of recovering RNA of all sizes (including microRNAs) High RNA yield directly from whole blood, no need to isolate WBCs
  TRIzol LS PureLink Total RNA Blood Kit PureLink Pro 96 total RNA Purification Kit MagMAX for Stabilized Blood Tubes RNA Isolation Kit MagMAX mirVana Total RNA Isolation RiboPure Blood Kit
Top Seller          
Compatible stabilization reagents EDTA, Heparin, Citrate, RNAlater EDTA, Heparin, Citrate EDTA, Heparin, Citrate Tempus or PaxGene tubes EDTA or Citrate EDTA, Heparin, Citrate, RNAlater
Stabilization reagent included          
RNAlater
Isolation method High purity organic extraction (requires alcohol precipitation) Fast, convenient silica column Fast, convenient silica columns in a plate format Scalable, flexible format with magnetic beads Scalable, flexible format with magnetic beads Highest purity & convenience; includes both organic extraction & silica column (no precipitation required)
Prep time 1 hour 20 minutes   12 tubes in 2 hours 96 samples in <1 hr 30 minutes
High throughput compatible      
For more info on working with blood please review our technical articles:
Supplemental Protocols
TechNotes Articles
Purifying RNA from cultured cells has traditionally been a highly repetitive, time- and labor-intensive procedure, often involving the use of toxic organic chemicals. The products developed by our RNA experts allow you to isolate RNA of maximum yield, purity, and integrity using several technology formats (organic extraction, glass-fiber columns, magnetic beads, and combinations of these).
You can also analyze cultured cell lysates without first isolating the RNA. In significantly less time, you’ll get qPCR results equivalent to - or in some cases better than - those from purified RNA. The Invitrogen products such as  Cells-to-CT and Single Cell-to-CT product lines offer tremendous performance and handling advantages.
Which RNA Isolation Kit from Cultured Cells is Right for You?
  Order now Order now Order now Order now Order now Order now
  Highly pure, intact RNA Fast and convenient column based format for RNA (>200 nt) Fast and convenient column based, filter plateformat for 96 samples (Rna > 200 nt) Fast and convenient column based method from limited sample input (Rna >200 nt) Fast and convenient method for small RNA species Complete system for cells to qRT-PCR without RNA purification
  TRIzol Reagent PureLink RNA Mini Kit PureLink Pro 96 total RNA Purification Kit PureLink RNA Micro Scale Kit MirVana miRNA Isolation Kit Cells-to-CT kits
Top seller    
Prep time 1 hour 20 minutes 20 minutes 20 minutes 30 minutes <10 minutes
Cell lysis method High purity organic extraction (requires alcohol precipi-tation) Nontoxic guanidine-isothio-cyanate lysis Nontoxic guanidine-isothio-cyanate lysis Nontoxic guanidine-isothio-cyanate lysis Organic extraction followed by alcohol precipitation Cells-to-CT lysis
Amount of Starting material Up to 1x107 cells 5x106 to 1x108 cells 5x106 to 1x108 cells Up to 5x105 cells 102-107 cultured cells 10-100,000 cells
Appli-cation All RNA expression methods qRT-PCR, NGS qRT-PCR, NGS qRT-PCR, NGS Micro RNA analysis qRT-PCR
Preserve and Stabilize Your RNA
RNAlater Solution is widely used for the preservation of RNA in intact animal tissues, organs, and cells, and bacteria. It eliminates the need to immediately process samples for RNA isolation or to freeze them in liquid nitrogen for later processing.
Technical Resources
Which RNA Extraction Kit from FFPE Samples is Right for You?
  Order now Order now
  Low-throughput isolation of RNA and DNA from the same FFPE section Scalable isolation of RNA and DNA from the same FFPE section
  RecoverAll Total Nucleic Acid Isolation Kit for FFPE MagMAX FFPE DNA/RNA Ultra Kit
Prep Time 4.5 hours (total time for 16 samples)
2.5 hrs hands-on time
4 hours (total time for 96 preps)
1 hr hands-on time
Final product extracted total RNA, microRNA, gDNA total RNA, microRNA, gDNA
Isolation method column purification mag bead purification
Deparaffinization required
Automation compatible (post deparffinization)  
(Scripts for KF Flex and Duo Prime)
Compatible FFPE sample range up to 4 x 20 micron sections
(300 mm2 tissue)
up to 2 x 20 micron sections
(300 mm2 tissue)
Sample volume ~50µL purified DNA
~50µL purified RNA
~50uL purified DNA
~50µL purified RNA
Compatible with Ion AmpliSeq DNA panels (e.g. Cancer Hotspot Panel v2)
Compatible with Ion AmpliSeq RNA panels (e.g. RNA cancer and RNA fusion)
Why is RNA Isolation From FFPE such a Challenge?
The recovery of quality RNA from FFPE specimens can be quite challenging. The fixation process causes cross-linkage between nucleic acids and proteins, and covalently modifies RNA by the addition of monomethyl groups to the bases. As a result, the molecules are rigid and susceptible to mechanical shearing, and the cross-links may compromise the use of RNA as a substrate for reverse transcription. Therefore, in order to utilize FFPE tissues as a source for gene expression analysis, a reliable method is required for extraction of RNA from the cross-linked matrix.
How can you Increase your Chances of Success With FFPE?
There are a number of factors that can impact the overall quality and yield of RNA isolated from FFPE tissues. Here are recommendations to address several key factors:
  1. Upstream tissue procurement and tissue specimen preparation—if possible, tissues should be fixed within one hour of surgical resection. Extensive degradation of RNA can occur before completion of the fixation process. The optimal fixation time is 12–24 hours, using neutral-buffered formalin or paraformaldehyde. Fixed tissues should be thoroughly dehydrated prior to the embedding process.
  2. Block storage—storage of blocks without cut faces, when possible, prevents ongoing damage from exposure to atmospheric oxygen, water, and other environmental factors such as light and infestation (fungus, insects, etc.).
  3. Tissue type, size, and amount being used for RNA isolation—the recommended tissue thickness is 10–20 µm The number of sections used is determined by the tissue type (which impacts cell density) and surface area (recommended size: 50–300 mm2). Excess starting material can cause filter clogging, resulting in poor yield.
  4. Excessive amount of paraffin used for embedding tissues—when possible, excess paraffin should be trimmed away prior to starting the purification protocol. For xylene-based purification methods, two xylene treatments at room temperature should be sufficient for complete deparaffinization. If desired, a more rigorous 37–55°C treatment can be performed for up to 30 minutes. After the xylene deparaffinization, it is crucial that the 100% ethanol is completely removed and the pellets are dry after the two 100% ethanol washes. The magnetic bead method employs novel chemistries to deal with the paraffin that limits input to 20 µm sections.
The MagMAX FFPE DNA/RNA Ultra Kits are designed for rapid, efficient, and automatable isolation of total RNA and DNA from FFPE samples using Invitrogen MagMAX magnetic particles. Invitrogen MagMAX FFPE NA kits offer faster workflows and use less toxic reagents, without sacrificing quality in the process. These kits help deliver nucleic acid yield and purity comparable to the best-in-class RecoverAll filter-based system. The MagMAX FFPE NA kits outperform products with similar protocols, and achieve RNA and DNA yields that are comparable with longer, more hazardous, and lower-throughput protocols.
The RecoverAll Total Nucleic Acid Isolation Kit procedure requires about 45 minutes of hands-on time and can easily be completed in less than 1 day when isolating RNA. FFPE samples are deparaffinized using a series of xylene and ethanol washes. Next, they are subjected to a rigorous protease digestion with an incubation time tailored for recovery of either RNA or DNA. The nucleic acids are purified using a rapid glass-filter methodology that includes an on-filter nuclease treatment, and are eluted into either water or the low-salt buffer provided.
Technical Resources
TechNotes Articles
Tips from the Bench
Your Data
Application Note
Product Bulletin
Which Plant RNA Isolation Kit is Right for You?
  Order now Order now Order now Order now
  Great for difficult samples (conifer tissue & seeds) Quick & easy to use Efficient recovery of microRNA & small RNA Rapid & fully automated
  Plant RNA Reagent PureLink RNA Mini Kit mirVana miRNA Isolation Kit MagMAX-96 Total RNA Isolation Kit
Top seller      
RNA types isolated large RNA molecules only large RNA molecules only Small & large RNA molecules large RNA molecules only
Prep time 60 minutes <20 minutes 30 minutes <45 minutes
Amount of starting material Up to 1 g <250 mg 0.5 to 250 mg Up to 10 mg
Isolation method High purity organic extraction (requires alcohol precipitation) Fast, convenient silica column Highest purity & convenience; includes organic extraction & silica column (no precipitation required) Scalable, flexible format with magnetic beads
High throughput compatible      
Compatible products   Ambion Plant RNA Isolation Aid
Removes common plant contaminants
Ambion Plant RNA Isolation Aid
Removes common plant contaminants
Ambion Plant RNA Isolation Aid
Removes common plant contaminants
Remove Polysaccharides
One of the major issues with plant RNA isolation is the presence of problematic biomolecules, such as polyphenolic compounds and polysaccharides. The Ambion Plant RNA Isolation Aid minimizes many of these contaminants with the addition of polyvinylpyrrolidone, a high molecular weight polymer that binds to polyphenolics and polysaccharides and a pre-extraction spin step.
Preserve and Stabilize Your RNA
RNAlater Solution is widely for the preservation of RNA in intact animal tissues, organs, and cells, and bacteria. It minimizes the need to immediately process samples for RNA isolation or to freeze them in liquid nitrogen for later processing.
Technical Resources
Which tissue RNA extraction kit is right for you?
  Order now Order now Order now Order now Order now Order now Order now
  Gold standard for highly pure, intact RNA Simple, reliable, rapid method Fast and convenient column based, filter plateformat for 96 samples (Rna > 200 nt) Fast and convenient column based method from limited sample input (Rna >200 nt) Fast and convenient method for small RNA species Highest purity TRIzol + column purification, no precipitation required HTP format, capable of recovering RNA of all sizes (including microRNAs)
  TRIzol Reagent PureLink RNA Mini Kit PureLink Pro 96 total RNA Purification Kit PureLink RNA Micro Scale Kit mirVana miRNA Isolation Kit TRIzol Plus RNA Purification System MagMAX mirVana Total RNA Isolation
Top Seller            
Isolation method High purity organic extraction (requires alcohol precipitation) Fast, convenient silica column Fast Convenient silica column in a filter plate format Fast Convenient micro silica columns Organic extraction combined with convenient silica columns Highest purity and convenience; includes both organic extraction and silica column (no precipitation required) Scalable, flexible format with magnetic beads
Prep time 1 hour 20 minutes 20 minutes 20 minutes 30 minutes 1 hour <1 hour
Amount of starting material Up to 100 mg 10 mg to 100 mg 10 cells to 10 mg up to 10 mg up to 250 mg tissue Up to 100 mg Up to 50 mg
High throughput compatible          
Prep size 100 preps 50 preps 96 preps 50 preps 96 preps 50 preps 96 preps
RNA Yields From Animal Tissues
RNA content can vary widely between tissues, cell types, physiological states, etc. This chart provides general guidelines for estimating RNA yields from a variety of cells and tissues.
Table 1. Typical total RNA yields from 1 mg of various tissues.
Preserve and Stabilize Your RNA
RNAlater Solution is widely used for the preservation of RNA in intact animal tissues, organs, and cells, and bacteria. It eliminates the need to immediately process samples for RNA isolation or to freeze them in liquid nitrogen for later processing.
Technical Resources
Which Yeast RNA Extraction Kit is Right for You?
  Order now Order now Order now Order now
  Rapid & fully automated Quick & easy to use Designed for rigid yeast cell walls while maintaining expression profiles Fast and convenient column based, filter plateformat for 96 samples (Rna > 200 nt)
  Platinum Taq MagMAX for Microarrays Total RNA Isolation Kit PureLink RNA Mini Kit RiboPure-Yeast Kit PureLink Pro 96 total RNA Purification Kit
Top seller      
Includes specialized beads for cell lysis No No Zirconia beads No
Isolation method Scalable, flexible format for highest purity; includes organic extraction and magnetic beads Fast, convenient silica column Highest purity and convenience; includes both organic extraction and silica column Fast, convenient silica column in 96 well filter plate format
Prep time <1 hour 20 minutes 90 minutes (includes DNase treatment) 20 minutes
Amount of starting material Up to 5 x 106 cells Up to 5 x 108 cells Up to 3 x 108 cells 1.8 mL of fresh, log-phase yeast cells (OD660=0.6–0.8)
High throughput compatible      
Kit size 96 preps 50 preps 50 preps 96 preps
Technical Resources
Unlike eukaryotic cells, yeast and many bacterial strains are protected by cell walls that can be difficult to disrupt. As a first step, effective cell lysis (typically via mechanical disruption or enzymatic pretreatment) is required prior to any RNA isolation procedure.
A range of products developed by the Ambion RNA experts will help you isolate high yields of pure, intact RNA from bacteria and yeast.
Which Bacterial RNA Extraction Kit is Right for You?
  Order now Order now Order now Order now Order now
  No lysis procedures Fast & easy, 20 min Fast and convenient silica column based , filter plate format HTP, 45 min Ideal for hard-to-lyse gram + bacteria
  TRIzol Max Bacterial RNA Isolation Kit w/TRIzol & Max Bacterial Enhancement Reagent PureLink RNA Mini Kit PureLink Pro 96 total RNA Purification Kit MagMAX Total Nucleic Acid Isolation Kit RiboPure-Bacteria Kit
Top Seller        
Includes specialized beads for cell lysis No No No Zirconia beads Zirconia beads
Isolation method High purity organic extraction (requires alcohol precipitation) Fast, convenient silica column Fast, convenient silica column in 96 well filter plate format Scalable, flexible format with magnetic beads Highest purity & convenience; includes organic extraction & silica column (no precipitation required)
Prep time 1 hour 20 minutes 20 minutes 45 minutes 1 hour
Amount of starting material Up to 1 x 108 cells Up to 1 x 109 cells Up to 1 x 109 cells Up to 0.3 g solid samples or 175 µl liquid samples Up to 109 cells
High throughput compatible      
Kit size 100 preps 50 preps 96 preps 100 preps 50 preps
Technical Resources
Viral analysis of biological (animal, insect, plant, fungal, bacterial) and environmental (water, air, food) samples is increasingly conducted using molecular testing strategies that require viral RNA or DNA from the samples being tested. Purification of viral RNA poses unique challenges for getting good recovery and detection sensitivity. Ambion RNA experts have developed RNA purification products that are optimized to provide maximum viral RNA yield, purity, and integrity from a broad range of sample types in several format options.
"Highly concentrated viral RNA from the PureLink Viral RNA/DNA Mini Kit provides a tool for more sensitive and delicate downstream analysis on viral genomes”
Dongning Wang, PhD, in the laboratory of Feng Gao, MD, Duke Human Vaccine Institute, Duke University
Product Specifications
  Most Suitable Downstream Application
Product Format Real-Time RT-PCR Microarray Analysis Sequen-cing Northern Blot Cloning
Total RNA
PureLink Viral RNA⁄DNA Mini Kit Chaotropic salt-based lysis, silica filter      
RNAqueous Kit Chaotropic salt-based lysis, silica filter    
MagMAX AI⁄ND Viral RNA Isolation Kit Chaotropic salt-based lysis, magnetic particles      
MagMAX-96 AI⁄ND Viral RNA Isolation Kit Chaotropic salt-based lysis, magnetic particles, 96-well processing, automation compatible      
DNA/RNA
PureLink Pro 96 Viral RNA⁄DNA Purification Kit Chaotropic salt-based lysis, silica filter, 96-well processing, automation compatible        
MagMAX Total Nucleic Acid Isolation Kit Chaotropic salt-based lysis, magnetic particles, 96-well processing, automation compatible      
MagMAX-96 Viral RNA Isolation Kit Chaotropic salt-based lysis, magnetic particles, 96-well processing, automation compatible      
Dynabeads SILANE Viral NA Chaotropic salt-based lysis, magnetic beads, automation compatible          
iPrep PureLink Virus Kit Chaotropic salt-based lysis, magnetic particles, automation compatible        
MagMAX Viral RNA Isolation Kit Chaotropic salt-based lysis, magnetic particles        
MagMAX-96 Viral RNA Isolation Kit (5 x 96 rxns) Chaotropic salt-based lysis, magnetic particles, 96-well processing, automation compatible        
MicroRNA
mirVana miRNA Isolation Kit Chaotropic salt-based lysis, silica filter        
mirVana miRNA Isolation Kit (without phenol - must be purchased separately) Chaotropic salt-based lysis, silica filter        
TRIzol LS Reagent Lysis in TRIzol reagent, one-step reagent  
mRNA
Dynabeads mRNA DIRECT Kit Chaotropic salt-based lysis, magnetic beads      
Dynabeads mRNA DIRECT Micro Kit Chaotropic salt-based lysis, magnetic beads