Viral RNA From Liquid & Cell-Free Samples
Viral analysis of biological (animal, insect, plant, fungal, bacterial) and environmental (water, air, food) samples is increasingly conducted using molecular testing strategies that require viral RNA or DNA from the samples being tested. Purification of viral RNA poses unique challenges for getting good recovery and detection sensitivity. Ambion® RNA experts have developed RNA purification products that are optimized to provide maximum viral RNA yield, purity, and integrity from a broad range of sample types in several format options.
“Highly concentrated viral RNA from the PureLink® Viral RNA/DNA Mini Kit provides a tool for more sensitive and delicate downstream analysis on viral genomes”
Dongning Wang, PhD, in the laboratory of Feng Gao, MD, Duke Human Vaccine Institute, Duke University
Product Specifications
  Most Suitable Downstream Application
Product Format Real-Time RT-PCR Microarray Analysis Sequencing Northern Blot Cloning
Total RNA
PureLink® Viral RNA⁄DNA Mini Kit Chaotropic salt-based lysis, silica filter      
RNAqueous® Kit Chaotropic salt-based lysis, silica filter    
MagMAX™ AI⁄ND Viral RNA Isolation Kit Chaotropic salt-based lysis, magnetic particles      
MagMAX™-96 AI⁄ND Viral RNA Isolation Kit Chaotropic salt-based lysis, magnetic particles, 96-well processing, automation compatible      
DNA/RNA
PureLink® Pro 96 Viral RNA⁄DNA Purification Kit Chaotropic salt-based lysis, silica filter, 96-well processing, automation compatible        
MagMAX™ Total Nucleic Acid Isolation Kit Chaotropic salt-based lysis, magnetic particles, 96-well processing, automation compatible      
MagMAX™-96 Viral RNA Isolation Kit Chaotropic salt-based lysis, magnetic particles, 96-well processing, automation compatible      
Dynabeads® SILANE Viral NA Chaotropic salt-based lysis, magnetic beads, automation compatible          
iPrep™ PureLink® Virus Kit Chaotropic salt-based lysis, magnetic particles, automation compatible        
MagMAX™ Viral RNA Isolation Kit Chaotropic salt-based lysis, magnetic particles        
MagMAX™-96 Viral RNA Isolation Kit (5 x 96 rxns) Chaotropic salt-based lysis, magnetic particles, 96-well processing, automation compatible        
MicroRNA
mirVana™ miRNA Isolation Kit Chaotropic salt-based lysis, silica filter        
mirVana™ miRNA Isolation Kit (without phenol - must be purchased separately) Chaotropic salt-based lysis, silica filter        
TRIzol® LS Reagent Lysis in TRIzol® reagent, one-step reagent  
mRNA
Dynabeads® mRNA DIRECT™ Kit Chaotropic salt-based lysis, magnetic beads        
Dynabeads® mRNA DIRECT™ Micro Kit Chaotropic salt-based lysis, magnetic beads      
Resources
Nucleic Acid Purification and Analysis Support Center
Find tips, troubleshooting help, and resources for your nucleic acid purification & analysis applications.
DNA Purification Brochure
RNA Isolation & Purification Brochure