Find the right Cells-to-CT kit for your needs
Cells-to-CT kits help accelerate RT-qPCR gene expression analysis by skipping RNA purification prior to amplification. Analyze RNA directly from cell lysates.
To use this tool, simply use the filters to help find the kit that best fits your research needs.
Filters
×
  • >
  • >
  • >
  • >
  • >
  • >
Kit image   Product name   Detection method   PCR method   Sample input   Sample type   Cell lysis time  
TaqMan
2-step
10–100,000 cells
Mammalian cells
5 min
TaqMan
2-step
10–100,000 cells
Mammalian cells, 3D cell culture
7 min
SYBR
2-step
10–100,000 cells
Mammalian cells
7 min
TaqMan
1-step
10–100,000 cells
Mammalian cells
7 min
SYBR
1-step
10–100,000 cells
Mammalian cells
7 min
TaqMan
2-step
1–10 cells
Mammalian cells
7 min
TaqMan
2-step
≤100,000 cells
Primary hepatocytes
10 min
For Research Use Only. Not for use in diagnostic procedures.