• Ready-to-transfect molecules that specifically inhibit endogenous miRNAs
  • Single-stranded, modified RNA design results in tight binding and efficient inhibition
  • Pre-designed inhibitors for all miRNAs listed in miRBase—custom design available
  • Available individually or in full sets for miRNA screening
  • Purified and ready to transfect
Search for Ambion® Anti-miR™ miRNA Inhibitors
Anti-miR™ miRNA Inhibitors
Ambion® Anti-miR™ miRNA Inhibitors are chemically modified, single stranded nucleic acids designed to specifically bind to and inhibit endogenous microRNA (miRNA) molecules. These ready-to-use inhibitors can be introduced into cells using transfection or electroporation parameters similar to those used for siRNAs, and enable detailed study of miRNA biological effects.
Use of the Anti-miR miRNA Inhibitors enable miRNA functional analysis by down-regulation of miRNA activity. Specific experimental designs include: miRNA target site identification and validation, screening for miRNAs that regulate the expression of a gene, and screening for miRNAs that affect a cellular process.
Anti-miR™ let-7c miRNA Inhibitor Positive Control
Anti-miR hsa-let-7c miRNA Inhibitor provides a convenient, validated positive control for experiments using Anti-miR miRNA Inhibitors. Endogenous let-7 miRNA negatively regulates HMGA2, a ubiquitously expressed, nonhistone chromatin protein that modulates gene expression through changes in chromatin architecture. When transfected into human and mouse cells, Anti-miR hsa-let-7c miRNA Inhibitor blocks endogenous let-7c miRNA, resulting in increased levels of HMGA2 mRNA. Thus, Anti-miR hsa-let-7c miRNA Inhibitor activity can be monitored in human or mouse cells by qRT-PCR using a TaqMan® Gene Expression Assay to HGMA2 (Assay IDs: Hs00171569_m1 [Human], Mm00780304_sH [Mouse]).
Anti-miR™ miRNA Inhibitor Negative Controls
Anti-miR Negative Control #1 is a random sequence Anti-miR molecule that has been extensively tested in human cell lines and tissues and validated to not produce identifiable effects on known miRNA function. Cy™3 and FAM™ dye-labeled Anti-miR Negative Controls are also available for monitoring transfection efficiency in transfection experiments using Anti-miR miRNA Inhibitors. The fluorescent label enables direct observation of the cellular uptake, distribution, and localization of the control. These dye-labeled controls are labeled at their 5' end and have the same oligonucleotide sequence as unlabeled Negative Control #1.
Anti-miR™ miRNA Inhibitor Libraries
The Anti-miR miRNA Inhibitor Library - Human consists of 1090 miRNA inhibitors corresponding to 1,240 human mature miRNAs cataloged in version 15 of the miRBase Sequence Database. This collection of miRNA inhibitors enables rapid study of miRNA function and can be used to simplify identification of miRNAs that interact with a particular target transcript. Each Anti-miR miRNA Inhibitor is supplied in a quantity of 0.25 nmol, dried in individual wells of a 96 well plate. This amount is sufficient for 50 transfections when used at 50 nM in 96 well plates. miRNA Precusor libraries plated as 1.0 nmol, dried in individual wells of a 96 well plate are also available.
Search More Than 2000 Ambion® miRNA Mimics and Inhibitors
Meet the Inventor: Silencer® Select siRNA technology
Search our more than 2000 Ambion® miRNA Mimics and Inhibitors 
Watch the video and learn how >