Thermo Fisher Scientific offers the most comprehensive miRNA collections available!
  • 100% coverage of miRBase Sequence Database Version 21 
  • Coverage to miRBase version 22 available as custom fulfillment
  • 96-well or 384-well plate format for high-throughput screening
  • Echo-qualified microplates for use with acoustic liquid handlers available upon request
  • Complete plate layout information and miRNA annotation provided
Learn more about our individual pre-designed mirVana Mimics & Inhibitors
Pre-defined mirVana miRNA mimic and inhibitor libraries
If you’re ready to start screening with a pre-defined collection or want to request a custom library, request a quote now!
Request a quote
Catalog #pre-defined collections for human and mouse miRNAnmol/well# of miRNA
4464074mirVana miRNA Mimics, Human v 21 - 384 or 96-Well Plates0.25 nmol2565
4464092mirVana miRNA Inhibitors, Human v 21 - 384 or 96-Well Plates0.25 nmol2565
4464075mirVana miRNA Mimics, Human v 21 - 384 or 96-Well Plates1 nmol2565
4464093mirVana miRNA Inhibitors, Human v 21 - 384 or 96-Well Plates1 nmol2565
4464074mirVana miRNA Mimics, Mouse v 21 - 384 or 96-Well Plates0.25 nmol1900
4464092mirVana miRNA Inhibitors, Mouse v 21 - 384 or 96-Well Plates0.25 nmol1900
4464075mirVana miRNA Mimics, Mouse v 21 - 384 or 96-Well Plates1 nmol1900
4464093mirVana miRNA Inhibitors, Mouse v 21 - 384 or 96-Well Plates1 nmol1900
Custom miRNA mimic and inhibitor libraries
In addition to our pre-defined genome-wide libraries, mirVana mimics and inhibitors are available as custom collections.
  • 96-well, 384-well, or Echo-qualified 384-well plates compatible with acoustic liquid handlers
  • Perfect for pilot studies, primary screen follow-up, or a customized functional genomics screen
  • Minimum order of 20 wells
Request a quote for a custom miRNA library
Product lines for custom library fulfillmentQuantity per well
mirVana miRNA mimics–human, mouse, or rat, miRBase v220.25 or 1.0 nmol/well
mirVana miRNA inhibitors–human, mouse, or rat, miRBase v220.25 or 1.0 nmol/well
Publications featuring mirVana miRNA libraries
The following selected peer-reviewed citations demonstrate the utility of mirVana mimic and inhibitor libraries in high-throughput screening applications to assess microRNA involvement in biological pathways such as those of development and growth, metabolic pathways and diseases, and cancer progression.
Lopes, IR et al. (2019) Shedding light on microRNA function via microscopy-based screening Methods 152: 55-64.
This comprehensive overview of miRNA mimic and inhibitor screening provides a detailed case study of the identification of miRNAs that control infection by the bacterial pathogen Salmonella. It includes methodologies that are readily adaptable for screens to address a wide variety of biological questions.
Takagawa Y, et al. (2020) miR-1293, a Candidate for miRNA-Based Cancer Therapeutics, Simultaneously Targets BRD4 and the DNA Repair Pathway Molecular Therapy 28(6): 1494-1505.
A genome-wide human mirVana miRNA mimic screen carried out in 10 cancer cell lines uncovered three tumor-suppressive microRNAs which target the gene BRD4, suggesting potential candidate for miRNA-based cancer therapeutics.
Meyer H-J, et al. (2017) Identification of a novel miRNA that increases transient protein expression in combination with valproic acid Biotechnology Progress 33(4): 1139-1145.
A custom collection of 875 mirVana mimics was screened to successfully identify two human miRNAs that increase recombinant protein titers for the purpose of large molecule therapeutic development.
Pal AS, et al. (2020) Identification of microRNAs that promote erlotinib resistance in non-small cell lung cancer Biochemical Pharmacology.https://doi.org/10.1016/j.bcp.2020.114154
Multiple microRNAs that drive EGFR-inhibitor resistance were identified in this genome-wide screen using mirVana miRNAs, uncovering a model for how miRNA dysregulation may contribute to drug resistance.
Resources
Ensure effective delivery, detection, and interpretation of your miRNA modulation experiments
Learn more about native regulatory RNAs
Support
If you have any questions on miRNA products or ordering, please contact us at RNAiSupport@thermofisher.com
For Research Use Only. Not for use in diagnostic procedures.