TaqMan® MicroRNA Assays & Arrays
TaqMan® MicroRNA Assays—Unrivaled sensitivity, specificity, and dynamic range
Our MicroRNA solutions feature TaqMan® MicroRNA Assay sensitivity, specificity, and dynamic range for unsurpassed miRNA quantitation. Our comprehensive Applied Biosystems solution provides the flexibility to meet different experimental needs and is supported by a complete range of reagents, assays, and real-time PCR instruments.
MicroRNA Assay & Array Workflows: Find the Products You Need for Every Step
MicroRNA Assay & Array Workflows: Find the Products You Need for Every Step
Get Same-Day Results with Megaplex™ Primer Pools and TaqMan® MicroRNA Arrays
- Megaplex™ Primer Pools simplify and accelerate miRNA profiling by reducing hands-on time and the amount of sample you need to as little as 1ng of total human, mouse or rat RNA—up to 100 times less material than existing analysis tools.
- Matched with the streamlined workflow enabled by TaqMan® MicroRNA Arrays, you can achieve reproducible RT-PCR data that delivers unrivaled sensitivity and specificity across a comprehensive set of miRNA targets derived from the Sanger database.
Also Available in 384-well Arrays
TaqMan® MicroRNA Arrays provide all the advantages of TaqMan® MicroRNA Assays in a convenient, pre-configured 384-well micro fluidic card, reducing experimental variability and the effort required to run 384 assays in parallel.
| Step 1: Extract Total RNA |
Step 2: Reverse Transcribe RNA |
Step 3: Set Up Singleplex TaqMan® MicroRNA Assay Reactions |
Step 4: Perform Real-Time PCR Amplification |
Step 5: Analyze Data |
|---|---|---|---|---|
|
||||
![]() |
![]() |
![]() |
![]() |
![]() |
| 30 min | 65 min | 25 min | 90 min |
| Step 1: Prepare Sample |
Step 2: Reverse Transcribe |
Step 3: Amplify cDNA (optional) |
Step 4: Dilute cDNA & Load TaqMan® Array |
Step 5: Run TaqMan® Array |
Step 6: Analyze Data |
|---|---|---|---|---|---|
|
|
|
|||
![]() |
![]() |
![]() |
![]() |
![]() |
![]() |
| 30 min | 150 min | 90 min | 10 min | 120 min |