BSC-1500B2
  • Meet the requirements of a Class II, Type B2 BSC under NSF/ANSI 49, a global standard for design, construction and performance of Class II biosafety cabinets (BSCs). 
  • Class II, Type B2 BSCs are often selected for applications for work with hazardous drugs, CMR (carcinogenic, mutagenic, reprotoxic) substances or other specialized procedures, particularly in North America.
  • NSF/ANSI 49 is the generally required standard for BSCs in North America as well as being frequently required and recommended in Latin America, the Asia Pacific, the Middle East and Africa. 
Select the class that suits your needs
1500 Series B2
Description Outstanding protection, comfort and performance for demanding applications.
Protection → SmartFlow
→ UV with saved cycle times
Ergonomics → Counterbalanced manual window
→ LCD information display located easily visible within sample chamber
→ Touch pad controls within reach while seated
Design → Stainless steel interior (Type 304)
→ Indented surface
Efficiency → DC intelligent supply motor
→ LED lights
1500 B2
The Thermo Scientific 1500 Series B2 BSC meets NSF/ANSI 49 for a Class II, Type B2 biosafety cabinet. These energy efficient cabinets require direct connection to external exhaust and provide personal and product protection from biological hazards and contamination as well as protection from volatile toxic chemicals used in the cabinet.
These cabinets are differentiated by:
  • SmartFlow compensation to maintain downflow allowing effective use of downflow prefilter for extended HEPA filter life.
  • Counter balanced window for easy operation.
  • 8 inch (20cm) front opening with Type 304 stainless steel interior.
Size/Width
4-foot (1.2m)
6-foot (1.8m)
Exterior dimensions (W x H x D) in 54.3 x 71.5 x 31.6 78 x 71.5 x 31.6
mm 1379 x 1816 x 803 1981 x 1816 x 803
Interior dimensions (W x H x D) in 48.5 x (25.7-29.2) x 25.5 72.5 x (25.7-29.2) x 25.5
mm 1232 x (653-742) x 648 1841 x (653-742) x 648
Shipping dimensions (W x H x D) in 72 x 81 x 44 86 x 81 x 44
mm 1829 x 2057 x 1118 2184 x 2057 x 1118
Working height of front window 8 inches (20.3cm) 8 inches (20.3cm)
Maximum opening height of front window
21.75 inches (55.2cm) 21.75 inches (55.2cm)
Damper 10 inch diameter air-tight damper included.
Size/Width
4-foot (1.2m)
6-foot (1.8m)
Net weight 545 lbs (247kg) 720 lbs (327kg)
Shipping weight1 646 lbs (293kg) 836 lbs (379kg)
Size/Width
4-foot (1.2m)
6-foot (1.8m)
Voltage (V) 115V or 230V 115V or 230V
Frequency (Hz) 50/60
Filter Specification 99.99% @ 0.3 micrometers
Certification NSF/ANSI 49, ETL, CE
Size/Width
4-foot (1.2m)
6-foot (1.8m)
Sound pressure level 65 66
Sound pressure level (dbA) per NSF/ANSI 49 91 fc (980 lx)
Receptacles 2 GFI duplex fo 115V, 2 single for 230V
Service Valves Up to 4 total (2 on each side wall)
Size/Width
4-foot (1.2m)
6-foot (1.8m)
Exhaust flow measured with traverse (Concurrent Balance Value) cfm 852 CFM @ 1.8 inches w.g. 1265 CFM @ 2.2 inches w.g.
cmh 1448 CMH @ 448 Pa 2149 CMH @ 548 Pa
Energy consumption, operating set point2 280 370
Heat Emission at 25°C ambient 0.022 0.024
Inflow air volume 283 cfm (481 m3/h) 414 cfm (703 m3/h)
Request a quote 1511 1561
* Clean filters, fans operating at nominal set point, interior lighting activated
* ETL tests the products to the following UL and Canadian standards: UL Standard 61010-1, CAN/CSA Standard C22.2 No. 1010.1. CE Mark applies only to 230V models.
Key features to help you achieve your next discovery
Based on over 40 years of engineering and design breakthroughs, our expertise is represented in key features that span our portfolio. Together these enhancements provide valuable contamination control technologies, and advanced design for highly critical applications, plus simplicity which allows you to spend more time pursuing your goals.
Thermo Scientific Class II, Type A2 BSCs use separate downflow and exhaust fans. SmartFlow compensation maintains inflow and the overall balance of inflow and downflow through real-time adjustment of the exhaust fan for constant flow and comparable adjustment to downflow. Thermo Scientific 2030i has SmartFlow Plus with fully implemented independent compensation to maintain constant inflow and constant downflow. 
All Thermo Scientific BSCs provide visual and audible alarms to warn of improper window position and unacceptable variation in airflow. Thermo Scientific 2030i has a unique interface that graphically depicts the alarm condition, text that advises of the specific concern and selectable audible tone to allow easy differentiation between window position and airflow problems.
Thermo Scientific BSCs allow a choice of internal surfaces including more economical partially coated interiors, type 304 stainless steel or type 316 stainless steel for greater chemical resistance. Our BSCs offer either double sided walls with negatively pressurized interstitial space for enhanced structure containment or transparent side panels for improved ergonomics. Work surfaces within the cabinet can be flat for easy cleaning or indented for containment of larger spills onto the work surface.
Thermo Scientific Class II, Type A2 BSCs use separate downflow and exhaust fans with DC electric motors. This combination of energy efficient DC motors with the fan based balance greatly reduces energy consumption compared to to traditional AC motor units with mechanical balancing dampers. 
Thermo Scientific BSCs allow a choice of traditional fluorescent lights for cabinet illumination or more energy efficient and sustainable LED lighting.