Cytotoxic safety cabinets
  • Meets the requirements of a safety cabinet for cytotoxic substances and other CMR drugs under DIN 12980 and EN 12469, the European standard for design, construction and performance of Class II biosafety cabinets
  • Cytostatic safety cabinets are often selected for applications for work with hazardous drugs, CMR (carcinogenic, mutagenic, reprotoxic) substances or other specialized procedures, particularly in Europe
  • DIN 12980, the German Standard for cytostatic safety cabinets and isolators, is frequently recommended and required in Europe and is the major standard directly addressing safety cabinets for work with cytotoxic substances and other CMR drugs
Browse our complete portfolio or let one of our experts help you make a selection.
Contact us
Maxisafe 2030i
Description Outstanding protection, comfort and performance for demanding cytostatic safety applications with intuitive interface and connectivity.
Protection → SmartFlow Plus
→ Two stage HEPA filtration with contained replacement of contaminated filters
→ UV Plus and cross beam
→ Full text messaging for alarms and alerts
Ergonomics → Motorized window with one button start up/shut down 
→ Transparent side walls
→ Individually selectable tones for alert and alarms
Design → Over the grill armrests
→ Stainless steel interior (Type 316)
→ Indented work surface
Efficiency → Dual DC intelligent motors
→ LED lights
→ Stand-by mode for further energy savings
Counterbalanced Manual Window
Maxisafe 2030i
The Thermo Scientific Maxisafe 2030i meets EN 12469 for a Class II biosafety cabinet. These energy efficient cabinets provide both personal and product protection from biological hazards and contamination and can be connected to external exhaust to provide protection for volatile toxic chemicals used in the cabinet.
These cabinets are differentiated by:
  • SmartFlow Plus compensation to independently maintain and monitor inflow and downflow.
  • Graphical User Interface with available one button start up/shut down and enhanced connectivity.
  • Motorized window for easy operation, transparent side walls and Type 316 stainless steel interior.
  • SmartClean Plus providing maximum access to the cabinet interior through hinged window movement.
Size/Width 0.9m (3 ft) 1.2m (4 ft) 1.5m (5 ft) 1.8m (6 ft)
Exterior dimensions
(W x H x D)
mm 1000 x 1536 x 870 1300 x 1536 x 870 1600 x 1536 x 870 1900 x 1536 x 870
in 39.4 x 60.5 x 34.3 51.2 x 60.5 x 34.3 63.0 x 60.5 x 34.3 74.8 x 60.5 x 34.3
Interior dimensions
(W x H x D)
mm 900 x 780 x 630 1200 x 780 x 630 1500 x 780 x 630 1800 x 780 x 630
in 35.4 x 30.7 x 24.8 47.2 x 30.7 x 24.8 59.1 x 30.7 x 24.8 70.9 x 30.7 x 24.8
Shipping dimensions1
(W x H x D)  
mm 1305 x 1750 x 970 1605 x 1750 x 970 1905 x 1750 x 970 2205 x 1750 x 970
in 51.4 x 68.9 x 38.2 63.2 x 68.9 x 38.2 75.0 x 68.9 x 38.2 86.8 x 68.9 x 38.2
Working height of front window 200mm (8 inches)
Maximum opening height of front window
773mm (30.4 inches)
1 Export packaging dimensions and weights upon request
Size/Width 0.9m (3 ft) 1.2m (4 ft) 1.5m (5 ft) 1.8m (6 ft)
Net weight kg 170 200 230 280
lbs 375 441 507 617
Shipping weight1 kg 200 235 275 330
lbs 441 518 606 728
Max load capacity of one-piece work surface 50kg (110 lbs)
Max load capacity of single segmented work surfaces (per 30cm segment) 25kg (55 lbs)
1 Export packaging dimensions and weight upon request
Size/Width 0.9m (3 ft) 1.2m (4 ft) 1.5m (5 ft) 1.8m (6 ft)
Voltage (V) 230, 115V NA option
Frequency (Hz)
50/60
Filter Specification H14 HEPA EN 1822, 99.995% @ MPPS  
Certification TUV TUV, LNE TUV TUV
Size/Width 0.9m (3 ft) 1.2m (4 ft) 1.5m (5 ft) 1.8m (6 ft)
Lighting power lux 1000 929 1500  1418
fc 93 86 139 132
Sound pressure level (dbA) per EN 12469 59 57 59 59
Receptacles 2 rear wall 4 rear wall
Service Valves
2 on each side
SmartPorts Two 76mm (3") plugged cable ports, one on each wall
Size/Width 0.9m (3 ft) 1.2m (4 ft) 1.5m (5 ft) 1.8m (6 ft)
Energy consumption, operating set point2 (W) 230 240 390 390
Energy consumption, Night Set-Back mode (W) 55 80
Heat output, operating mode (non-vented) (BTU/hr) 785 819 1331 1331
Heat output, night set-back mode (lights off, non vented) (BTU/hr) 188 273
Exhaust / Inflow air volume (m3/h) 296 395 493 592
(cfm) 174 232 290 348
Exhaust volume, thimble ducted (m3/hr) 385 513 641 770
(cfm) 226 302 377 453
Request a quote 51032715 51032716 51032717 51032718
2 Clean filters, fans operating at nominal set point, interior lighting activated