NSF 49 Certified Biological Safety Cabinets
Protection that never takes a day off
Designed to minimize costs of operation and maintenance and maximize comfort and convenience for advanced ease of use, Class II, Type A2 biological safety cabinets provide best-in-class safety, ergonomics, and energy efficiency.
Special product features include:
- H14 HEPA filters increase filtration efficiency at 99.995% of MPPS
- Solid plenums for long life and lasting containment
- Negative-pressure, dual side walls protect from leaks to the exterior environment
Browse our complete portfolio or let one of our experts help you make a selection.
Save time, energy, and money with Thermo Scientific™ 1300 Series Class II, Type A2 Biological Safety Cabinets.
Perform daily applications safely and efficiently with this simple-to-order package, which includes a cabinet, stand, pre-installed UV light, and armrests.
Protect workers and samples during work with gases and volatile chemicals. Thermo Scientific™ 1300 Series Class II, Type B2 Biological Safety Cabinets are designed to meet the stringent safety requirements of working with toxic chemicals and trace amounts of radionuclides required in microbiological studies.
Optimize configuration flexibility of 1300 Series A2 Class II Biological Safety Cabinets with these optional stands, UV-lighting, arm rests, foot rests, valves, and piping.
Conduct quick, simple cleaning and fumigations with the Thermo Scientific™ Herasafe™ KS (NSF) Class II, Type A2 Biological Safety Cabinet, featuring an advanced design.
Unity Lab Services: Get access to our expert service team, information on support plans, parts and consumables resources, and other instrument services.