Brochure downloads
Designed to achieve your next breakthrough
The Thermo Scientific Heracell VIOS series represents a new era in advanced incubator design for sensitive cultures—like stem and primary cells—in leading research, pharmaceutical, and clinical applications.
Through a holistic approach to culturing, our newest incubator series provides everything necessary for your most demanding and highly critical applications. By combining our latest technology advances in contamination control and uniform growth conditions with existing proven and reliable features, you are now able to achieve your goals faster, more reliably, and with less effort.
  • Protected humidity reservoir 
  • In situ sensors and probes 
  • Steri-Run high-temp decontamination 
  • Thermo Scientific iCAN intuitive touchscreen
  • Direct heat chamber
  • Thermo Scientific THRIVE active airflow 
  • ISO 5 HEPA air filtration
Featured Heracell VIOS CO2 incubators
stainless steel chamber
The Thermo Scientific Heracell VIOS 160i CO2 Incubator offers revolutionary THRIVE active airflow technology, delivering homogeneous growth conditions fast while avoiding unwanted sample variation. 
100% pure copper chamber
The Thermo Scientific Heracell VIOS 160i CO2 Incubator provides protection from every direction, including ISO class 5 HEPA filtered air, on-demand high temperature sterilization, and an easy-to-maintain 100% copper chamber. 
Tri-Gas incubator
The Thermo Scientific Heracell VIOS 160i Tri-Gas Incubator is designed to focus on convenience, allowing you to spend more time on your research and less time managing your incubator. 
Featured video
See THRIVE active airflow in action
Thermo Scientific THRIVE active airflow technology provides faster recovery and uniformity for consistent results. Cells experience total recovery of all critical growth parameters in less than 10 minutes following a door opening.