Introducing new Forma CO2 Incubators
Discovery thrives in a culture of confidence
More scientists worldwide trust their valuable cultures to Thermo Scientific™ CO2 incubators than any other brand. The inside story is simple: our CO2 incubators let you culture with confidence. Day after day. Year after year.
Now with the intuitive Thermo Scientific™ iCAN™ touch screen interface, the Thermo Scientific™ Forma™ Series 3 Water-Jacketed CO2 Incubator provides reliable temperature stability through its unique triple-wall construction. It also offers ISO 5 HEPA filtration for continuous clean room air quality.
Forma Steri-Cycle i160 Direct Heat CO2 Incubators
The Thermo Scientific™ Forma™ Steri-Cycle™ i160 series represents a new era in advanced incubator design for sensitive cultures—like stem and primary cells—in leading research, pharmaceutical, and clinical applications.
Performance and protection for every day culturing. Available with ISO Class 5 HEPA filtration system, which is designed to provide continuous protection against unwanted airborne contaminants; 140°C on-demand high temperature sterilization cycle that simplifies routine cleaning; and CO2 control with choice of TC or IR sensors.
The optimal choice for GMP, bioproduction or large scale culturing of high value samples, the Thermo Scientific™ Forma™ Steri-Cult CO2 Incubator brings leading edge technology into your lab.
Outstanding value for every day culturing, the stackable Thermo Scientific™ Forma™ direct heat incubator with optional HEPA filter airflow system, provides a large capacity and is easy-to-maintain.
Learn more about the first choice of researchers around the world because of the reliability of proven water jacket technology. Durable triple-wall construction delivers optimal temperature uniformity. And it’s guaranteed for life against leakage. It provides outstanding thermal stability for your valuable cell cultures to protect against ambient temperature swings and unexpected power outages that can ruin your cultures.
View brochure ›