FMS High-Performance Lab Freezers
High-performance cold storage
Protect flammable materials, solvents and intermediates using Thermo Scientific™ TSHP FMS High-Performance Laboratory Freezers. All freezers meet stringent U.S. Pharmacopeia standards for drug storage as well as the NFPA safety provisions for the storage of flammable liquids.
Find your model ›
Popular FMS high-performance lab freezers
FMS High-Performance Laboratory Freezers
FMS High-Performance Laboratory Freezers
Designed to meet stringent US Pharmacopeia standards for drug storage as well as the NFPA safety provisions for the storage of flammable liquids, our new FMS high-performance laboratory refrigerators and freezers feature:
  • Easy-to-use, intuitive digital display featuring microprocessor control
  • Lockable doors, audio and visual alarms including door ajar alarms for security and peace of mind
  • Convenient access ports and caster
  • Ignition-free interior cabinets for the safe storage of flammable materials
  • Manual defrost