-20 & -30 Lab Freezers
Manual defrost -20°C and automatic defrost -30°C laboratory freezers accommodate a variety of capacities, demanding, space challenged environments, and samples and applications ranging from routine to complex.
  • Environmentally-friendly, non CFC and HCFC refrigerants
  • Energy savings without sacrificing protection
  • Alarms built in, plus connectivity options for state-of-the-art monitoring and reporting
  • Quiet operation
  • Keyed security, including power switch and setpoint
  • Easy mobility plus lockable casters
  • Ergonomic handles
  • GMP Clean Room Class A / ISO 6 (ISO EN 14644-1) compatible (TSX series only)
Browse our complete portfolio or contact one of our experts to help you make a selection.
Automatic defrost freezers, when compared to manual defrost freezers, do not require you to remove samples prior to defrosting, saving you time to focus on what matters most: your work.
Automatic defrost freezers feature forced air circulation for uniform temperature throughout the cabinet, provide faster door opening recovery, and feature non-invasive temperature and time based defrost cycles.  
Sample types: Reagents, pharmaceuticals, biologicals, and other commonly used laboratory materials such as primers, siRNA molecules, and fetal bovine serum
  • V-drive technology provides improved temperature uniformity that continually adapts to the lab or clinical enviroment 
  • Whisper quiet operation 
  • GMP Clean Room Class A / ISO 6 (ISO EN 14644-1) compatible with appropriate pre-install preparation
Value
General Purpose
High Performance
Budget-friendly lab storage for routine samples.Designed for medical and scientific applications and featuring large, easy-to-read icon-based controls and displays, microprocessor control and scratch-resistant cold-rolled steel interior and exterior finishes.Designed with V-drive technology that supports critical sample protection and sustainability objectives, and provides whisper quiet operation for clinical and laboratory-grade storage.
GP Series Lab Freezer ›(115v/60Hz only)
GPS Series Lab Freezer › (Not available in North America or other 60Hz regions)
Revco HP Freezer ›
Jewett HP Freezer ›
Forma HP Freezer ›
TSG Series Freezers ›
TSX HP Freezer ›
Learn more about auto defrost freezers ›
Manual defrost freezers, or cold wall freezers, are most commonly used in BioTech and Pharma R&D applications, and are designed for applications in which even slight cabinet warming during auto defrost cannot be tolerated.
Manual defrost freezers contain refrigeration coils located inside the walls of the cabinet. The coils transfer refrigerant to the walls of the cabinet, whereas auto defrost freezers utilize forced air circulation and heat injection to prevent frost build-up.   
Sample types: Enzymes, pharmaceuticals, MMR vaccines, and other samples that require -20°C storage
  • V-drive technology provides improved temperature uniformity that continually adapts to the lab or clinical enviroment 
  • Whisper quiet operation 
  • GMP Clean Room Class A / ISO 6 (ISO EN 14644-1) compatible with appropriate pre-install preparation
Learn more about manual defrost freezers ›
Related products
Plasma Freezers
erlenmeyer-flasks-170x170
Related resources
Brochures:
TSX Series High-Performance Refrigerators
and Freezers
TSX Undercounter Refrigerators
ENERGY STAR Infographic