Enjoy a wide selection of ULT storage products and many great choices depending on your particular needs.  We’ve made it easy to find the right freezer by selecting from the following category selections.
Selection guide
-80 Benchtop
-80 Chest
-80 TDE/RDE/FDE Upright
-40 TDE/RDE/FDE Upright
-80 TSX Upright
Description
Small footprint, benchtop ULT freezer ideal for personal use.
Chest style floor models intended for dependable, long term sample storage where infrequent sample access is the norm.
The workhorse of the busy lab, these upright models set the standard for performance and reliable, high-capacity sample storage in heavy use situations, whether you require -80 or -40 capabilities.
Feature rich, premium performance for busy, modern labs. Built for the most critical sample storage and challenging lab environments, with outstanding energy efficiency.
Temperature Capability
-50C to -80C -50C to -80C
-50C to -80C
-10C to -40C
-50C to -80C
Storage Capacity 2” Boxes 16 63-396 300-600 300-600 400-700
Storage Capacity Volume
1 cubic foot (28L)
3 cubic feet (84.9 L) – 20 cubic feet (566.3 L)
14.9 cubic feet (422 L) – 28.8 cubic feet (816 L)
14.9 cubic feet (422 L) – 28.8 cubic feet (816 L)
19.4 cubic feet (549 L) – 33.5 cubic feet (949 L)
Compliance
General Purpose
General Purpose
GP/FDA Class II
GP/FDA Class II
General Purpose
Sound
Normal
Normal
Quiet
Quiet
Extremely Quiet
Voltage Capability
115V/60Hz
208-230V/60Hz
115V/60Hz
208-230V/60Hz
 230V/50Hz
115V/60Hz
208-230V/60Hz
 230V/50Hz
115V/60Hz
208-230V/60Hz
 230V/50Hz
115V/60Hz
208-230V/60Hz
 230V/50Hz
Energy Star Rated
Select 600 2" Box Models
Select 600 2" Box Models
yes
Energy Consumption
As low as 5.3 kWh/day
As low as 11kWh/day
As low as 10.5 kWh/day
As low as 8kWh/day
as low as 7.9kWh/day
System Refrigerants
HFC
HFC
Natural HC, environmentally friendly
Natural HC, environmentally friendly
Natural HC, environmentally friendly
User Interface
Push Button LED
HIC Touch Button
HIC Touch Button
VIC Touchscreen
Cloud Connect Enabled
No
No
No
No
Yes
Available Options
CO2/LN2 Backup
CO2/LN2 Backup
Chart Recorder
CO2/LN2 Backup Chart Recorder
CO2/LN2 Backup
Chart Recorder
CO2/LN2 Backup
HID Access Control
Chart Recorder
Explore all models
Once you have found the category of ULT freezers that meets your needs, you can explore different models and select the one that is ideal for your lab.