standard-value-lab-refrigerators-278x195
An economical, safe storage solution for basic or routine laboratory applications requiring temperatures from 1° to 12°C and that benefit from adjustable temperature control and manual defrost. These refrigerators are not suitable for high value life science materials or clinical biologics, nor are they suitable for corrosive environments.
  • Warranty of 13 months parts and labor
  • Each unit is factory tested prior to shipment
  • Environmentally-friendly, non CFC refrigerants
  • Easy-to-clean, durable finish
Browse our complete portfolio or contact one of our experts to help you make a selection.
Choose a value lab refrigerator
Undercounter general purpose refrigerator for indoor storage of non-critical samples and reagents (ADA compliant model available in North America).
Undercounter Value Lab Refrigerators
ADA-Compliant Undercounter Value Lab Refrigerators
Undercounter Value Lab Refrigerators
  General purpose refrigerator for indoor storage of non-critical samples and reagents. (North America only)
ADA-compliant general purpose refrigerator for indoor storage of non-critical samples and reagents. (North America only) General purpose refrigerator for indoor storage of non-critical samples and reagents. (Outside North America only)
Request a quote
Capacity 5.5 cu. ft. (155.7L) 5.5 cu. ft. (155.7L) 5.5 cu. ft. (155.7L)
Temperature Range 1° to 12°C 1° to 12°C 1° to 12°C
Doors 1 1 1
Shelves 3 4 3
Interior (DxWxH) 17.5 x 20 x 27.75 in. (44.5 x 50.8 x 70.5cm) 17.5 x 20 x 27.75 in. (44.5 x 50.8 x 70.5cm) 17.5 x 20 x 27.75 in. (44.5 x 50.8 x 70.5cm)
Exterior (DxWxH) 23.5 x 23.63 x 33.5 in. (60 x 60 x 85.1cm) 24.25 x 23.65 x 32 in. 23.5 x 23.63 x 33.5 in. (60 x 60 x 85.1cm)
Shipping Weight 100 lb. (45.3kg) 102 lb. (46.3kg) 100 lb. (45.3kg)
Upright general purpose refrigerator for indoor storage of non-critical samples and reagents.
Upright Value Lab Refrigerators
  General purpose refrigerator for indoor storage of non-critical samples and reagents (North America only).
Request a quote
Capacity 20 cu. ft. (566.34L)
Temperature Range 1° to 12°C
Doors 1
Shelves 4
Interior (DxWxH) 23.8 x 27.8 x 57.8 in. (60.3 x 70.5 x 146.7cm)
Exterior (DxWxH) 32.13 x 34 x 73.13 in. (81.6 x 86.4 x 186.1cm)
Shipping Weight 283 lb. (128kg)
Undercounter general purpose refrigerator for indoor storage of non-critical samples and reagents (ADA compliant model available in North America).
Undercounter Value Lab Refrigerators
ADA-Compliant Undercounter Value Lab Refrigerators
Undercounter Value Lab Refrigerators
  General purpose refrigerator for indoor storage of non-critical samples and reagents. (North America only)
ADA-compliant general purpose refrigerator for indoor storage of non-critical samples and reagents. (North America only) General purpose refrigerator for indoor storage of non-critical samples and reagents. (Outside North America only)
Request a quote
Capacity 5.5 cu. ft. (155.7L) 5.5 cu. ft. (155.7L) 5.5 cu. ft. (155.7L)
Temperature Range 1° to 12°C 1° to 12°C 1° to 12°C
Doors 1 1 1
Shelves 3 4 3
Interior (DxWxH) 17.5 x 20 x 27.75 in. (44.5 x 50.8 x 70.5cm) 17.5 x 20 x 27.75 in. (44.5 x 50.8 x 70.5cm) 17.5 x 20 x 27.75 in. (44.5 x 50.8 x 70.5cm)
Exterior (DxWxH) 23.5 x 23.63 x 33.5 in. (60 x 60 x 85.1cm) 24.25 x 23.65 x 32 in. 23.5 x 23.63 x 33.5 in. (60 x 60 x 85.1cm)
Shipping Weight 100 lb. (45.3kg) 102 lb. (46.3kg) 100 lb. (45.3kg)
Upright general purpose refrigerator for indoor storage of non-critical samples and reagents.
Upright Value Lab Refrigerators
  General purpose refrigerator for indoor storage of non-critical samples and reagents (North America only).
Request a quote
Capacity 20 cu. ft. (566.34L)
Temperature Range 1° to 12°C
Doors 1
Shelves 4
Interior (DxWxH) 23.8 x 27.8 x 57.8 in. (60.3 x 70.5 x 146.7cm)
Exterior (DxWxH) 32.13 x 34 x 73.13 in. (81.6 x 86.4 x 186.1cm)
Shipping Weight 283 lb. (128kg)
Top
Which plug type do I need for my value refrigerator?
  Standard Plug Type Voltage Hz
NEMA 5-15 115–120 60
NEMA 5-20 115–120 60
NEMA 6-15 208–230 60
Continental EU
230 50
Featured Brochure
Value-LR-F_60Hz_1217
Value Lab Refrigerators, Freezers and Combination Units
An economical solution for safe sample storage.
Open brochure
Related products
vaccine-storage-170x170
fms-freezer-170x170
deivce-link-hub-170x170
erlenmeyer-flasks-170x170
Related resources
Brochures:
TSX Series High-Performance Refrigerators
and Freezers
TSX Undercounter Refrigerators
ENERGY STAR Infographic