upright-medical-refrigerators-freezers-278x195
When you need clinical-grade storage it's important to choose a refrigerator or freezer that is designed specifically for the demands that come with these kinds of environments. Whether it's a concern with temperature variation impacting sensitive materials, size, sound, energy use, Clean Room compliance, or all of the above and more, we have the storage solution for you and for the needs of your environment.
Browse our complete portfolio or contact one of our experts to help you make a selection.
Choose an upright medical refrigerator by application
Blood Bank Refrigerators
Chromatography Refrigerators
Pharmacy Refrigerators
Our high-performance blood bank refrigerators are designed to meet strict requirements established by the AABB for the storage of whole blood and blood components.TSX chromatography refrigerators provide our best temperature control, full access to chromatography insturmentation, and feature industry-leading safety features to ensure maximum protection.Our high-performance pharmacy refrigerators feature adjustable basket drawers and glass doors for easy identification and access to vaccines, pharmaceuticals and chemotherapeutics.
  • TSX V-drive technology offers the greater sample integrity, reduced noise, and energy savings
  • Easy-to-access controls with setpoint security
  • Blood bank refrigerators and plasma freezers meet strict guidelines for storing whole blood and blood components
  • V-drive technology adapts to your activity to stabilize temperatures and save energy
  • Internal electrical outlet protected by safety sensor circuit
  • Two side access ports to accommodate chromatography applications and equipment
  • GMP Clean Room Class A / ISO 6 (ISO EN 14644-1) compatible
  • Featuring V-drive technology
  • Roll-out wire basket drawers
  • GMP Clean Room Class A / ISO 6 compatible
Choose an upright medical freezer by application
Enzyme Freezers
Plasma Freezers
Support sample protection and sustainability objectives for the storage of enzymes requiring –20°C where even slight intermittent coil warming during auto defrost cannot be tolerated.Our high-performance –30°C plasma freezers with automatic defrost are designed to meet strict requirements established by the AABB for the storage of plasma.
  • Featuring V-drive technology
  • Comes standard with 9 shelves and bins for convenient enzyme storage
  • GMP Clean Room Class A / ISO 6 compatible
  • Featuring V-drive technology
  • Comes standard with 7 stainless steel rollout drawers for convenient plasma bag storage
  • GMP Clean Room Class A / ISO 6 compatible
Featured Brochure
TSX505 Undercounter Ref Bro-270x195
TSX Series high-performance undercounter refrigerators
The only undercounter lab-grade refrigerator with redundant cooling.
  • Low energy usage
  • Low noise
  • High storage capacity
Related products
storage
Vaccine Storage Solutions
Wireless Monitoring Solutions
Reusable Plasticware
Related resources
Brochures:
TSX Series High-Performance Refrigerators
and Freezers
TSX Undercounter Refrigerators
ENERGY STAR Infographic