When storing blood and plasma it's important to choose a refrigerator or freezer that is designed specifically for the demands of blood and plasma storage. Whether it's a concern with temperature variation impacting sensitive materials, size, sound, energy use, Clean Room compliance, or all of the above and more, we have the blood and plasma storage solutions for your blood bank.
- TSX V-drive technology adapts to user activity to stabilize temperatures and save energy
- Meet strict requirements established by the AABB for storage of whole blood and blood components
- TSX provides redundant refrigeration systems that provide ultimate sample security
- Cutting edge innovation to meet demands of regulated environments
Browse our complete portfolio or contact one of our experts to help you make a selection.
Quick links
Choose a blood bank refrigerator
Choose a single door blood bank refrigerator
11.5 cu.ft TSX Series Blood Bank Refrigerator |
23 cu. ft. TSX Series Blood Bank Refrigerator |
29.2 cu.ft. TSX Series Blood Bank Refrigerator |
|
![]() |
![]() |
![]() |
|
Energy Star certified and designed with features that support AABB requirements for the storage of whole blood and blood components. | |||
Request a quote |
|||
Capacity | 11.5 cu .ft. (326L) | 23 cu. ft. (650L) | 29.2 cu. ft. (827L) |
Temperature Range | 2° to 8°C | 2° to 8°C | 2° to 8°C |
Doors | Single, Glass | Single, Glass | Single, Glass |
Interior (DxWxH) | 21.8 x 20.0 x 52.4 in. (55.4 x 50.8 x 133.1cm) | 28.5 x 24 x 58 in. (72.3 x 61 x 147.3cm) | 28.5 x 30.0 x 58.0 in. (72.3 x 76.2 x 147.3cm) |
Exterior (DxWxH) | 30.5 x 24.0 x 73.0 in. (77.5 x 61.9 x 185.4cm) | 37.8 x 28 x 78.6 in. (96 x 71.1 x 199.6cm) | 37.3 x 34.0 x 78.5 in. (94.7 x 86.4 x 199.4cm) |
Shipping Weight | 404 lb. (183kg) | 526 lb. (239kg) | 554 lb. (251kg) |
11.5 cu. ft. Forma Blood Bank Refrigerators |
29.2 cu. ft. Forma Blood Bank Refrigerators |
|
![]() |
![]() |
|
Meet strict requirements for storage of whole blood and blood components with microprocessor control, simplified push-button alarm test functions and reliable, field-proven refrigeration. |
||
Request a quote |
||
Capacity | 11.5 cu .ft. (326L) | 29.2 cu. ft. (827L) |
Temperature Range | 1° to 4°C | 1° to 4°C |
Doors | Single, Glass | Single, Glass |
Interior (DxWxH) | 20.5 x 20 x 52.4 in. (50.8 x 50.8 x 133.1cm) | 29 x 30 x 58 in. (73.7 x 76.2 x 147.3cm) |
Exterior (DxWxH) | 29.3 x 24 x 73.6 in. (74.3 x 61 x 186.9cm) | 36.4 x 34 x 79.2 in. (92.5 x 86.4 x 201.2cm) |
Shipping Weight | 220 lb. (96kg) | 505 lb. (227kg) |
Single, Glass |
11.5 cu. ft. Revco Blood Bank Refrigerators |
29.2 cu. ft. Revco Blood Bank Refrigerators |
|
![]() |
![]() |
|
Store whole blood and blood components while meeting strict requirements, featuring microprocessor control systems, positive airflow systems, industrial-quality cabinet construction and extra-strength refrigeration. |
||
Request a quote |
||
Capacity | 11.5 cu.ft. (326L) | 29.2 cu. ft. (827L) |
Temperature Range | 1° to 8°C | 1° to 8°C |
Doors | Single, Glass | Single, Glass |
Interior (DxWxH) | 21.75 x 20 x 52.4 in. (55.2 x 50.8 x 133.1cm) | 29 x 30 x 58 in. (73.7 x 76.2 x 147.3cm) |
Exterior (DxWxH) | 29.3 x 24 x 73.6 in. (74.3 x 61 x 186.9cm) | 36.4 x 34 x 79.2 in. (92.5 x 86.4 x 201.2cm) |
Shipping Weight | 320 lb. (145kg) | 505 lb. (229kg) |
11.5 cu. ft. Jewett Blood Bank Refrigerators |
29.2 cu. ft. Jewett Blood Bank Refrigerators |
|
![]() |
![]() |
|
Store whole blood and blood components while meeting strict requirements, featuring microprocessor control systems, positive airflow systems, industrial-quality cabinet construction and extra-strength refrigeration. | ||
Request a quote |
||
Capacity | 115 cu. ft. (326L) | 29.2 cu. ft. |
Temperature Range | 1° to 8°C | 1° to 8°C |
Doors | Single, Glass | Single, Glass |
Interior (DxWxH) | 21.75 x 20 x 52.4 in. (55.2 x 50.8 x 133.1cm) | 29 x 30 x 58 in. (73.7 x 76.2 x 147.3cm) |
Exterior (DxWxH) | 29.3 x 30 x 73.6 in. (74.3 x 76.2 x 186.9cm) | 36.4 x 34 x 79.2 in. (92.5 x 86.4 x 201.2cm) |
Shipping Weight | 320 lb. (145kg) | 505 lb. (227kg) |
Single, Glass |
Single, Glass |
Choose a double door blood bank refrigerator
51.1 cu. ft. TSX Series Blood Bank Refrigerators |
|
![]() |
|
Energy Star™ certified and designed with features that support sample protection and sustainability objectives for the storage of whole blood and blood components. | |
Request a quote |
|
Capacity | 51.1 cu. ft. (1447L) |
Temperature Range | 2° to 8°C |
Doors | Double, Glass |
Interior (DxWxH) | 28.5 x 52.5 x 58 in. (72.4 x 133.4 x 147.3cm) |
Exterior (DxWxH) | 37.8 x 56.5 x 78.5 in. (96 x 143.5 x 199.4cm) |
Shipping Weight | 865 lb. (392kg) |
Featured Brochure
TSX Series High-Performance Refrigerators and Freezers
Lab and clinical-grade storage that adapts to you and your environment.
Featured Resource
Green fact sheet: TSX Series high-performance freezers
This fact sheet provides the rationale behind the environmental claim that Thermo Scientific™ TSX Series high performance freezers, including large format models, meet ENERGY STAR™ criteria for lab-grade refrigerators and freezers.
Related products
Related resources
Brochures:
TSX Series High-Performance Refrigerators
and Freezers
TSX Undercounter Refrigerators
ENERGY STAR Infographic