Designed for a variety of applications requiring close temperature control, full access to chromatography instrumentation, and easy set-up of instrumentation and apparatus within the chamber.
- Industry-leading safety features to ensure maximum protection
- Forced-air circulation for temperature uniformity at all shelf levels
- Heat-free defrost for maximum temperature uniformity
Browse our complete portfolio or contact one of our experts to help you make a selection.
Quick links
Choose a chromatography refrigerator
TSX Series Chromatography Refrigerators |
||||
![]() |
||||
Industry-leading safety features to ensure maximum protection for chromatography applications requiring a 2° to 8°C controlled environment, with full access to chromatography instrumentation and easy set-up within the chamber. | ||||
Request a quote | ||||
Capacity | 23 cu. ft. (650L) | 29 cu. ft. (827L) | 45.8 cu. ft. (1297L) | 51.1 cu. ft. (1447L) |
Temperature Range | 2° to 8°C | 2° to 8°C | 2° to 8°C | 2° to 8°C |
Doors | Single, Glass | Single, Glass | Sliding Glass | Double, Glass |
Interior (DxWxH) | 28.5 x 24 x 58 in. (72.3 x 61 x 147.3cm) | 28.5 x 30.0 x 58.0 in. (72.3 x 76.2 x 147.3cm) | 26 x 52.5 x 58 in. (66 x 133.4 x 147.3cm) | 28.5 x 52.5 x 59 in. (72.4 x 133.4 x 147.3cm) |
Exterior (DxWxH) | 37.8 x 28 x 78.5 in. (96.2 x 71.1 x 199.4cm) | 37.3 x 34.0 x 78.5 in. (94.7 x 86.4 x 199.4cm) | 36.6 x 56.5 x 78.5 in. (93.1 x 143.5 x 199.4cm) | 37.8 x 56.5 x 78.5 in. (96 x 143.5 x 199.4cm) |
Shipping Weight | 439 lb. (199kg) | 442 lb. (210kg) | 600 lb. (272kg) | 658 lb. (298kg) |
Forma Chromatography Refrigerators |
||
![]() |
||
Excellent temperature stability and full accessibility with microprocessor control system, positive airflow systems, industrial-quality cabinet construction and extra-strength refrigeration compressors. | ||
Request a quote | ||
Capacity | 29 cu. ft. (827L) | 78.8 cu. ft. (2231L) |
Temperature Range | 1° to 8°C | 1° to 8°C |
Doors | Single, Glass | Triple, Glass |
Interior (DxWxH) | 29 x 30 x 58 in. (73.7 x 76.2 x 147.3cm) | 29 x 30 x 58 in. (73.7 x 76.2 x 147.3cm) |
Exterior (DxWxH) | 36.4 x 34 x 79.2 in. (92.5 x 86.4 x 201.2cm) | 36.4 x 34 x 79.2 in. (92.5 x 86.4 x 201.2cm) |
Shipping Weight | 435 lb. (197kg) | 435 lb. (197kg) |
Revco Chromatography Refrigerators |
||
![]() |
||
Precise temperature control with positive airflow systems, industrial-quality cabinet construction and extra-strength refrigeration compressors, plus access ports with hinged covers for full access to instrumentation. | ||
Request a quote | ||
Capacity | 29.2 cu. ft. (827L) | 78.8 cu. ft. (2231L) |
Temperature Range |
1° to 8°C | 1° to 8°C |
Doors | Single, Glass | Triple, Glass |
Interior (DxWxH) | 29 x 30 x 58 in. (73.7 x 76.2 x 147.3cm) | 29 x 81 x 58 in. (73.7 x 205.7 x 147.3cm) |
Exterior (DxWxH) | 29 x 34 x 79.2 in. (73.7 x 86.4 x 201.2cm) | 29 x 85 x 79.2 in. (73.7 x 215.9 x 201.2cm) |
Shipping Weight | 435 lb. (197kg) | 965 lb. (438kg) |
TSG Chromatography Refrigerators |
||||
![]() |
||||
Built with performance, security and quality features required of non-critical products and samples that need a 2° to 10°C controlled environment, plus safety sensor protected interior outlet and a whisper-quiet operation. | ||||
Request a quote |
||||
Capacity | 27 cu. ft. (764L) | 45. cu. ft. (1274L) | 49 cu. ft. (1387L) | 72 cu. ft. (2038L) |
Temperature Range | 2˚ to 10˚C | 2˚ to 10˚C | 2˚ to 10˚C | 2˚ to 10˚C |
Doors | Single, Glass | Double, Glass | Double, Glass | Triple, Glass |
Interior (DxWxH) | 28.5 x 27 x 61.75 in. (724 x 686 x 1569mm) | 27 x 49 x 61.75 in. (686 x 1245 x 1569mm) | 28.5 x 49 x 61.75 in.(724 x 1245 x 1569mm) | 28.5 x 72 x 61.75 in. (724 x 1829 x 1569mm) |
Exterior (DxWxH) | 32 x 30.75 x 81 in. (813 x 781 x 813mm) | 32 x 52.7 x 81 in. (813 x 1339 x 813mm) | 32 x 52.7 x 81 in. (813 x 1339 x 813mm) | 32 x 75.7 x 81 in (813 x 1924 x 813mm) |
Shipping Weight | 386 lb. (175kg) | 561 lb. (255kg) | 597 lb. (271kg) | 733 lb. (332kg) |
Featured Brochures
TSX Series High-Performance Refrigerators and Freezers
Lab and clinical-grade storage that adapts to you and your environment.
TSG Series General Purpose and Chromatography Refrigerators
TSG Series Chromatography Refrigerators are designed with your chromatography applications in mind.
Related products
Related resources
Brochures:
TSX Series High-Performance Refrigerators
and Freezers
TSX Undercounter Refrigerators
ENERGY STAR Infographic