upright-enzyme-freezer-278x195
Our enzyme freezers are –20°C manual-defrost upright freezers designed for applications in which even slight intermittent coil warming during auto defrost cannot be tolerated.
  • ENERGY STAR® certified (TSX series only)
  • Enzyme bins included for convenient storage
  • Cold wall convection cooling for temperature uniformity at all shelf levels or gravity convection air systems without blowers or fans
  • Easy mobility with four 2"casters, front two are lockable
Browse our complete portfolio or contact one of our experts to help you make a selection.
Choose an enzyme freezer
TSX Enzyme Freezers
 High-performance -20°C manual defrost enzyme freezers designed to support sample protection and sustainability objective with V-drive technology to provide ultimate temperature uniformity that continually adapts to user patterns, offering significant energy savings without compromising protection.
Request a quote
Capacity23 cu. ft. (650L)29.2 cu. ft. (827L)
Temperature Range−35° to −15°C-25° to -15°C
ConnectivityNASmart-Vue Compatible
DoorsSingle, SolidSingle, Solid
Interior (DxWxH)28.5 x 24 x 58 in. (72.3 x 61 x 147.3cm)28.5 x 30.0 x 58.0 in. (72.3 x 76.2 x 147.3cm)
Exterior (DxWxH)37.5 x 28 x 78.6 in. (95.3 x 71.1 x 199.6cm)38.9 x 34.0 x 78.5 in. (99.0 x 86.4 x 199.4cm)
Shipping Weight476 lb. (216kg)552 lb. (251kg)
Looking for additional Enzyme Freezer Storage Bins?
Related products
vaccine-storage-170x170
fms-freezer-170x170
deivce-link-hub-170x170
erlenmeyer-flasks-170x170
Related resources
Brochures:
TSX Series High-Performance Refrigerators
and Freezers
TSX Undercounter Refrigerators
ENERGY STAR Infographic