upright-pharmacy-refrigerator-278x195
Our high-performance pharmacy refrigerators feature adjustable basket drawers and glass doors for the easy identification and access of vaccines, pharmaceuticals, chemotherapeutics and other material requiring 2° to 8°C storage.
  • ENERGY STAR® certified (TSX series only)
  • Triple panel glass door helps maintain optimal internal conditions
  • Forced-air circulation for temperature uniformity and fast temperature recovery
  • Heat-free defrost for maximum temperature uniformity
Browse our complete portfolio or contact one of our experts to help you make a selection.
Choose a pharmacy refrigerator
TSX Series Pharmacy Refrigerators
  The quiet, sustainable choice for medical and pharmacy-grade refrigeration, designed to provide temperature uniformity that continually adapts to user patterns, offering significant energy savings without compromising protection.
Request a quote
Capacity 11.5 cu. ft. (326L) 23 cu. ft. (650L) 29.2 cu. ft. (827L) 51.1 cu. ft. (1447L)
Temperature Range 2° to 8°C 2° to 8°C 2° to 8°C 2° to 8°C
Connectivity Smart-Vue Compatible DLH compatible Smart-Vue Compatible DLH compatible
Doors Single, Glass Single, Glass Single, Glass Double, Glass
Interior (DxWxH) 21.8 x 20.0 x 52.4 in. (55.4 x 50.8 x 133.1cm) 28.5 x 24 x 58 in. (72.3 x 61 x 147.3cm) 28.5 x 30.0 x 58.0 in. (72.3 x 76.2 x 147.3cm) 28.5 x 52.5 x 58 in. (72.4 x 133.4 x 147.3cm)
Exterior (DxWxH) 30.5 x 24.0 x 73.0 in. (77.5 x 61.9 x 185.4cm) 37.8 x 28 x 78.6 in. (96 x 71.1 x 199.6cm) 37.3 x 34.0 x 78.5 in. (94.7 x 86.4 x 199.4cm) 37.9 x 56.5 x 78.5 in. (96.2 x 143.5 x 199.4cm)
Shipping Weight 388 lb. (176kg) 510 lb. (231kg) 541 lb. (245kg) 830 lb. (376kg)
Forma Pharmacy Refrigerators
  Store and retrieve pharmacy products with ease, developed for stable temperature control over a range of 1° to 8°C, featuring microprocessor control, positive airflow systems and industrial-quality cabinet construction.
Request a quote
Capacity 11.5 cu. ft. (326L) 29.2 cu. ft. (827L)
Temperature Range 1° to 8°C 1° to 8°C
Doors Single, Glass Single, Glass
Interior (DxWxH) 21.75 x 20 x 52.4 in. (55.2 x 50 x 132cm) 29 x 30 x 58 in. (73.7 x 76.2 x 147.3cm)
Exterior (DxWxH) 29.3 x 24 x 73.6 in. (74.3 x 61 x 186.9cm) 36.4 x 34.0 x 79.2 in. (92.5 x 86.4 x 201.2cm)
Shipping Weight 320 lb. (145kg) 505 lb. (227kg)
Revco Pharmacy Refrigerators
  Simplify operations and increase accuracy with pharmacy refrigerators designed for stable temperature control over a range of 1° to 8°C plus positive airflow systems, industrial-quality cabinet construction and extra-strength compressors.
Request a quote
Capacity 11.5 cu. ft. (326L) 29.2 cu. ft. (827L)
Temperature Range 1° to 8°C 1° to 8°C
Doors Single, Glass Single, Glass
Interior (DxWxH) 21.8 x 20 x 52.4 in. (53.3 x 50.8 x 133.1cm) 29 x 30 x 58 in. (73.3 x 76.2 x 147.3cm)
Exterior (DxWxH) 29.3 x 24 x 73.6 in. (74.3 x 61 x 186.9cm) 36.4 x 34.0 x 79.2 in. (92.5 x 86.4 x 201.2cm)
Shipping Weight 320 lb. (145kg) 505 lb. (227kg)
Jewett Pharmacy Refrigerators
  Securely store and easily retreive pharmacy products with these refrigerators developed for stable temperature control 1° to 8°C featuring stainless-steel, roll-out drawers and standard safety glass double-pane doors.
Request a quote
Capacity 29.2 cu. ft. (827L)
Temperature Range 1° to 8°C
Doors Single, Glass
Interior (DxWxH) 29 x 30 x 58 in. (73.7 x 76.2 x 147.3cm)
Exterior (DxWxH) 36.4 x 34 x 79.2 in. (92.5 x 86.4 x 201.2cm)
Shipping Weight 505 lb. (227kg)
Featured Brochure
TSX series brochure
TSX Series High-Performance Refrigerators and Freezers
Lab and clinical-grade storage that adapts to you and your environment.
Open brochure
Featured Resource
TSX series brochure
Green fact sheet: TSX Series high-performance freezers
This fact sheet provides the rationale behind the environmental claim that Thermo Scientific™ TSX Series high performance freezers, including large format models, meet ENERGY STAR™ criteria for lab-grade refrigerators and freezers.
Open fact sheet
Related products
vaccine-storage-170x170
fms-freezer-170x170
deivce-link-hub-170x170
erlenmeyer-flasks-170x170
Related resources
Brochures:
TSX Series High-Performance Refrigerators
and Freezers
TSX Undercounter Refrigerators
ENERGY STAR Infographic