upright-plasma-freezer-278x195
Maintain storage integrity while ensuring fast heat removal and temperature recovery. Designed to meet AABB, ANRC, and FDA standards for safety and performance. Our plasma freezers are ideal for demanding blood bank applications.
  • Forced-air circulation for temperature uniformity and fast temperature
  • Heat-free defrost for maximum temperature uniformity
  • Convenient, stainless steel, fully extendable and adjustable drawers
  • Easy mobility with four 2"casters, front two are lockable
Browse our complete portfolio or contact one of our experts to help you make a selection.
Choose a plasma freezer
TSX Plasma Freezers
 Designed with features that support sample protection and sustainability objectives for the storage of plasma. The Series’ V-drive technology is designed to provide temperature uniformity that continually adapts to user patterns, offering significant energy savings without compromising protection.
Request a quote
Capacity11.5 cu. ft. (326L)23 cu. ft. (650L)29.2 cu. ft. (827L)51.1 cu. ft. (1447L)
Temperature Range−35° to −15°C−35° to −15°C−35° to −15°C−35° to −15°C
DoorsSingle, SolidSingle, SolidSingle, SolidDouble, Solid
Interior (DxWxH)21.8 x 20.0 x 52.4 in. (55.4 x 50.8 x 133.1cm)28.5 x 24.0 x 58.0 in. (72.3 x 61 x 147.3cm)28.5 x 30.0 x 58.0 in. (72.3 x 76.2 x 147.3cm)28.5 x 52.5 x 58 in. (72.4 x 133.4 x 147.3cm)
Exterior (DxWxH)31.1 x 24.0 x 73.0 in. (79.0 x 61.9 x 185.4cm)37.9 x 28 x 78.6 in. (96.2 x 71.1 x 199.6cm)37.9 x 34.0 x 78.5 in. (96.2 x 86.4 x 199.4cm)37.8 x 56.5 x 78.5 in. (96 x 143.5 x 199.4cm)
Shipping Weight412 lb. (187kg)526 lb. (239kg)556 lb. (252kg)817 lb. (371kg)
Forma Plasma Freezers
 Manage demanding blood bank applications with microprocessor-controlled plasma freezers designed for simplified operation and greater accuracy. These −30°C freezers meet AABB, ANRC and FDA standards for safety and performance in plasma storage.
Request a quote
Capacity11.5 cu. ft. (326L)29.2 cu. ft. (827L)
Temperature Range−30°C−30°C
DoorsSingle, SolidSingle, Solid
Interior (DxWxH)21 x 20 x 52.4 in. (53.3 x 50.8 x 133.1cm)29 x 30 x 58 in. (73.7 x 76.2 x 147.3cm)
Exterior (DxWxH)29.3 x 24 x 73.6 in. (74.3 x 61 x 186.9cm)37.2 x 34 x 79.2 in. (94.5 x 86.4 x 201.2cm)
Shipping Weight365 lb. (145kg)580 lb. (263kg)
Single, Solid
Revco Plasma Freezers
 Simplify operation and increase accuracy with the microprocessor control our plasma freezers designed to meet AABB, ANRC and FDA standards for safety and performance in plasma storage.  
Request a quote
Capacity11.5 cu. ft. (326L)29.2 cu. ft. (827L)
Temperature Range−30°C−30°C
DoorsSingle, SolidSingle, Solid
Interior (DxWxH)21 x 20 x 52.4 in. (53.3 x 50.8 x 133.1cm)29 x 30 x 58 in. (73.7 x 76.2 x 147.3cm)
Exterior (DxWxH)29.3 x 24 x 73.6 in. (74.3 x 61 x 186.9cm)37.2 x 34 x 79.2 in. (94.5 x 86.4 x 201.2cm)
Shipping Weight365 lb. (166kg)580 lb. (263kg)
Jewett Plasma Freezers
 Meet the high standards for sensitive plasma freezing with multiple sizes that meet large and small storage requirements and feature easy-to-use microprocessor controls.
Request a quote
Capacity11.5 cu. ft. (326L)29.2 cu. ft. (827L)
Temperature Range−30°C−30°C
DoorsSingle, SolidSingle, Solid
Interior (DxWxH)20 x 21 x 52.4 in. (53.3 x 50.8 x 133.1cm)29 x 30 x 58 in. (73.7 x 76.2 x 147.3cm)
Exterior (DxWxH)29.3 x 24 x 52.4 in. (74.3 x 61 x 186.9cm)37.2 x 34 x 79.2 in. (94.5 x 86.4 x 201.2cm)
Shipping Weight365 lb. (145kg)580 lb. (263kg)
Related products
vaccine-storage-170x170
fms-freezer-170x170
deivce-link-hub-170x170
erlenmeyer-flasks-170x170
Related resources
Brochures:
TSX Series High-Performance Refrigerators
and Freezers
TSX Undercounter Refrigerators
ENERGY STAR Infographic