Search Thermo Fisher Scientific
Thermo Scientific versatile dry baths / block heaters offer a wide range of blocks to accommodate various tubes and plates.
Available with digital or touch screen controls, our units provide outstanding temperature uniformity and stability that help create the reproducible results that you need.
Touch Screen Dry Baths | Digital Dry Baths | Compact Dry Baths | Heating / Shaking Dry Baths | Heating / Cooling Dry Baths | |
---|---|---|---|---|---|
![]() | ![]() | ![]() | ![]() | ![]() | |
Function | Heating | Heating | Heating | Heating / shaking | Heating / cooling |
Block capacity | 1, 2, 4 | 1, 2, 4 | Dry bath S: 1-4 Dry bath D: 1-8 | 1 | 1 |
Max 1.5mL tube capacity | 112 | 112 | Dry bath S: 24 Dry bath D: 48 | 24 | 30 |
Controller | PID digital | PID digital | Digital | PID digital | PID digital |
Temp range | Amb +5° to 130°C (Amb @25°C) | Amb +5° to 130°C (Amb @25°C) | Amb +2° to 100°C (Amb @25°C) | Amb +5° to 100°C | 4 to 95°C |
Temp accuracy | ≤ ± 0.5°C | ≤ ± 0.5°C | ≤ ± 0.5°C | 0.1°C | 0.1°C |
Temp uniformity | ≤ ± 1°C | ≤ ± 1°C | ≤ ± 0.5°C | 0.55°C | 0.55°C |
Timer | 0-99:59 min or continuous | 0-99:59 min or continuous | NA | 1-99:59 min | 1-99:59 min |
Programmable | Programmable | NA | NA | Programmable | Programmable |
Temp probe | PT1000 | NA | NA | NA | NA |
Shaking speed | NA | NA | NA | 150 - 1,500 rpm | NA |
Warranty | 2 yrs | 2 yrs | 2 yrs | 2 yrs | 2 yrs |
Complete your purchase with a set of dry bath blocks. Available in a number configurations to support any application you desire. Use our simple tool to make block selection easier!