Search
Multi-point hot plate stirrers are ideal for high-volume, high-performance processing and life science/biotechnology applications as well as labs with multiple users.
Find the hot plate stirrer to meet your needs using our hot plate stirrer selection guide ›
| Super-Nuova Multi-Position Digital Stirring Hotplates | ||
|---|---|---|
![]() | ||
| Perform life science and biotechnology applications with the help of Thermo Scientific Super-Nuova Multi-Position Digital Stirring Hotplates, which offer four independently-controlled stirring positions on one heated, ceramic top. Including 4-function memory and a low-temperature adjustment ideal for life science and biotechnology applications, these stirrers are designed for labs with multiple users that require high-volume processing with unmatched performance, safety and controls. | ||
| Part no. | SP135935Q | SP135930-33Q |
| Plate material | Ceramic | Ceramic |
| Plate shape | Square | Square |
| Plate size | 10.25 x 10.25 in. (26.0 x 26.0cm) | 10.25 x 10.25 in. (26.0 x 26.0cm) |
| Voltage | 120V | 220/240V |
| Speed | 50 to 1,200rpm | 50 to 1,200rpm |
| Temperature Range | 1° to 370°C | 1° to 370°C |
| Stirring Points | 4 | 4 |
Unity Lab Services: Access to our expert service team, information on support plans, parts and consumables resources, and other instrument services.