Superspeed Rotors with Auto-Lock
Provides application flexibility for years to come
Offering up to 100,605 x g performance and 6 L capacity along with Thermo Scientific™ Fiberlite™ LEX rotor designs, the extensive rotor portfolio for the Thermo Scientific Sorvall™ LYNX™ superspeed centrifuge series supports labware—from microplates to microtubes and 1 L bottles—all with our push-button Thermo Scientific™ Auto-Lock™ rotor exchange.
Popular superspeed rotors with auto-lock
Featured superspeed rotor with auto-lock categories
Auto-Lock rotor exchange with Auto-ID instant rotor identification innovations simplify run set-up and help to eliminate overspeeding or rotor accidents on Sorvall LYNX superspeed centrifuges. Productivity features include speed handles on rotor lids that simplify lid tightening or removal. You can quickly swap between your sample preparation protocols in only 3 seconds with just the push of a button.
Our line of next-generation high-speed centrifuges, the Thermo Scientific Sorvall LYNX Superspeed Centrifuge Series supports multiple users and a variety of processing requirements, providing a range of user experience and flexible solutions to the everyday challenges of a shared laboratory setting.
Featured superspeed centrifugation videos