Ultracentrifuge Rotors
Simplicity and versatility
Our versatile ultracentrifuge rotor portfolio addresses your high-performance processing needs, accommodating tubes from 0.2 mL to 250 mL—and providing application flexibility as your lab’s research focus evolves. 
Our ultra rotor portfolio provides:
  • Fast pelleting and density gradient separations with fixed-angle rotors, including lightweight and durable Thermo Scientific™ Fiberlite™ carbon fiber rotors which improve ergonomics, backed by a 15-year warranty
  • High-performance swinging-bucket rotors for density gradient runs and for locating small pellets at the bottom of a tube
  • Fast lipoprotein and plasmid DNA separations with StepSaver™  vertical rotors that streamline sample loading and unloading
  • Continuous flow rotors ideal for multi-liter processing applications
  • Batch processing gradient applications with zonal rotor
Popular ultracentrifuge rotors
Featured ultracentrifuge rotor categories
Maximize space in your lab with the small footprint of the Thermo Scientific Sorvall™ WX+ Ultracentrifuge Series, while maintaining up to 100,000 rpm speed required for your high-performance processing applications.
Download brochure ›
Support
  • Unity Lab Services 
    Access to our expert service team, information on support plans, parts and consumables resources, and other instrument services.