Heraeus Centrifuges
Thermo Scientific Heraeus centrifuges and their innovative rotors meet your processing needs—spanning protein and cell biology to molecular biology and drug discovery to clinical research, and supporting labware from microplates and microtubes to large-capacity bottles. Heraeus products are designed to deliver outstanding performance, spin after spin.
Product selection guide to help you choose your Heraeus solution
Utilize our product selection tool to quickly and easily find your centrifuge and rotor combination.
Discover the Heraeus centrifuge portfolio
Accelerate your micro-volume separations with our compact, safe, and easy-to-use microcentrifuges, offering power and versatility for routine laboratory work as well as high-end applications. Available in two speed options with both refrigerated and ventilated models, including ready-to-use configurations, find the reliability, flexibility and convenience you need to maximize your lab's productivity.
Heraeus Pico and Fresco 17 and 21 Microcentrifuges
- 17,000 / 21,100 x g max RCF
- 13,300 / 14,800 rpm max speed
- 7 available rotors for 0.2–5mL tube volumes, spin columns, PCR strips, and capillary tubes
Learn more:
Thermo Scientific compact and general purpose centrifuges provide exceptional capacity (up to 4 liters) in compact designs with a smart, simple interface. Featuring innovative Thermo Scientific technologies such as Auto‐Lock rotor exchange and ClickSeal biocontainment lids, Fiberlite rotors and the flexibility to adapt to evolving clinical and research applications.
- 4 x 145mL (with TX-150 rotor), 6 x 50mL (with HIGHConic III rotor)
- Maximum g-force up to 30,279 x g
- Compact footprint
- Ventilated and refrigerated options
- 13 available rotors
- 1.6-liter capacity (4 x 400mL)
- Simplified routine operating interface
- Mid-performance drive
- Ventilated and refrigerated options
- 12 available rotors
- 1.6-liter capacity (4 x 400mL)
- Advanced research operating interface
- High-performance drive
- Ventilated and refrigerated options
- 14 available rotors
- 4-liter capacity (4 x 1,000mL)
- Simplified routine operating interface
- Mid-performance drive
- Ventilated and refrigerated options
- 10 available rotors
- 4-liter capacity (4 x 1,000mL)
- Advanced research operating interface
- High-performance drive
- Ventilated, refrigerated and floor models
- 17 available rotors
Introducing the Heraeus Cryofuge 8 and 16 centrifuges designed for flexible, large capacity blood banking. Enhanced ergonomics, glove-friendly touchscreen interface for quick run set-up and standardization, and improved capacity maximizes your lab's productivity.
- 16 x 500mL max blood bag capacity with available configurations supporting 6, 8, 12, or 16 bags
- 7,295 x g max RCF
- 5,000 rpm max speed
- Touchscreen interface supporting up to 120 programs
- Mobile app connectivity for setup and monitoring via our Centri-Vue application
Accelerate your micro-volume separations with our compact, safe, and easy-to-use microcentrifuges, offering power and versatility for routine laboratory work as well as high-end applications. Available in two speed options with both refrigerated and ventilated models, including ready-to-use configurations, find the reliability, flexibility and convenience you need to maximize your lab's productivity.
Heraeus Pico and Fresco 17 and 21 Microcentrifuges
- 17,000 / 21,100 x g max RCF
- 13,300 / 14,800 rpm max speed
- 7 available rotors for 0.2–5mL tube volumes, spin columns, PCR strips, and capillary tubes
Learn more:
Thermo Scientific compact and general purpose centrifuges provide exceptional capacity (up to 4 liters) in compact designs with a smart, simple interface. Featuring innovative Thermo Scientific technologies such as Auto‐Lock rotor exchange and ClickSeal biocontainment lids, Fiberlite rotors and the flexibility to adapt to evolving clinical and research applications.
- 4 x 145mL (with TX-150 rotor), 6 x 50mL (with HIGHConic III rotor)
- Maximum g-force up to 30,279 x g
- Compact footprint
- Ventilated and refrigerated options
- 13 available rotors
- 1.6-liter capacity (4 x 400mL)
- Simplified routine operating interface
- Mid-performance drive
- Ventilated and refrigerated options
- 12 available rotors
- 1.6-liter capacity (4 x 400mL)
- Advanced research operating interface
- High-performance drive
- Ventilated and refrigerated options
- 14 available rotors
- 4-liter capacity (4 x 1,000mL)
- Simplified routine operating interface
- Mid-performance drive
- Ventilated and refrigerated options
- 10 available rotors
- 4-liter capacity (4 x 1,000mL)
- Advanced research operating interface
- High-performance drive
- Ventilated, refrigerated and floor models
- 17 available rotors
Introducing the Heraeus Cryofuge 8 and 16 centrifuges designed for flexible, large capacity blood banking. Enhanced ergonomics, glove-friendly touchscreen interface for quick run set-up and standardization, and improved capacity maximizes your lab's productivity.
- 16 x 500mL max blood bag capacity with available configurations supporting 6, 8, 12, or 16 bags
- 7,295 x g max RCF
- 5,000 rpm max speed
- Touchscreen interface supporting up to 120 programs
- Mobile app connectivity for setup and monitoring via our Centri-Vue application
Looking to perform high-speed separations in a shared lab setting? Check out our high speed centrifuges and ultracentrifuges.
Heraeus centrifuge applications and techniques
Advance your research with proven cell culture instruments and equipment, including CO₂ incubators, biological safety cabinets, general purpose centrifuges, lab refrigerators and freezers, automated cell counters, and cell imaging systems.
Maximize your upstream and downstream productivity through enhanced harvest and purification techniques. Our centrifuges for bioprocessing are designed for high quality with run-to-run reproducibility and traceability for all your bioseparation needs, ranging from R&D scale to large-scale production.
Designed for solid reliability and quiet operation, our comprehensive range of clinical solutions (certified for in vitro diagnostic use) fit in your lab with their compact footprints with throughput from 1.5mL to 50mL sample tubes, microplates and 10mL syringes.