mySpin mini centrifuge
Mini centrifuge rotors to maximize speed and flexibility
Thermo Scientific mySPIN 12 and 6 mini centrifuge rotors enable fast spins up to 12,500 rpm and are ideal for microfilter cell separations and HPLC samples, with adapters to accommodate all major microtube and PCR tube styles (2.0, 1.5, 0.5, 0.2mL) for customized use.
Browse our complete portfolio or let one of our experts help you make a selection.
Contact us
Bring increased efficiency and user comfort to your lab
The mySPIN 12 and 6 mini centrifuges, providing 12- or 6-place rotors with tool-free, quick rotor exchange for ease-of-use, are adaptable for microfilter cell separation and HPLC protocols as well as PCR, clinical and diagnostic applications. Spin 0.2mL and 1.5/2.0mL tubes with mySPIN centrifuge rotors, and ensure continued, uninterrupted operation with mySPIN centrifuge replacement parts.
Related mini centrifuge rotors categories
mini centrifuges
microcentrifuges
centrifuge ware
Protein purification and isolation
Support
Unity Lab Services 
Access to our expert service team, information on support plans, parts and consumables resources, and other instrument services.