mini centrifuges
Mini centrifuges designed for your personal comfort and efficiency
Thermo Scientific mySPIN 12 and 6 mini centrifuges are designed for quick and pulse spins and offer convenience at your personal workstation. Tool-free rotors, duplicate spin function, and ergonomic design promotes both efficiency and ease of use. Safety features help ensure they will not operate when the lid is open.
Browse our mini centrifuges and resources below or let one of our product experts help you make a selection.
Contact us
Quick links
Adaptable for microfilter and HPLC protocols as well as PCR applications
Both mySPIN mini centrifuges are ideal for microfilter cell separation and HPLC protocols, with the mySPIN 12 centrifuge also supporting PCR, as well as clinical and diagnostic applications.
Both mini centrifuges offer:
  • Small footprint for efficient use of lab space
  • Tool-free, quick rotor exchange
  • Quiet operation, low vibration
  • Includes two rotors to accommodate 0.2, 0.5, 1.5, and 2.0mL microtubes and PCR strip tubes
Which mini centrifuge is right for you?
  mySPIN mySPIN
  mySPIN 6
mySPIN 12
Catalog # 75004061 75004081
Max RCF 2,000 xg (not adjustable) 9,800 xg (100 xg increments)
Max speed 6,300 xg (not adjustable) 12,500 xg (100 rpm increments)
Max capacity 6 x 2mL 12 x 2mL
Rotors & adapters included
  • 6 x 1.5/2.0mL circular rotor
  • 16 x 0.2mL rotor (singles or 2 x 8 strip)
  • 0.2mL tube adapters (set of 6)
  • 0.5mL tube adapters (set of 6)
  • 12 x 1.5/2.0mL rotor
  • 32 x 0.2mL rotor (singles or 4 x 8 strip)
  • 0.2mL tube adapters (set of 12)
  • 0.5mL tube adapters (set of 12)
Max noise level 51–53 dBA 51–53 dBA
Available timer settings NA 1 sec to 30 min (1 sec increments), and quick-spin
Dimensions
(H x W x D)
4.1 x 5.0 x 6.0in (10.4 x 12.8 x 15.3cm) 4.4 x 6.7 x 8.0in (11.4 x 17.1 x 20.3cm)
Weight 0.74 kg (1.6 lb) 1.41 kg (3.1 lb)
Certifications
  • CSA certified
  • CE marked
  • UL listed
mini centrifuge rotors
microcentrifuges
centrifuge ware
Protein purification and isolation
Support
Unity Lab Services 
Access to our expert service team, information on support plans, parts and consumables resources, and other instrument services.