Search Thermo Fisher Scientific
Combined technical data sheets and drawings are available in PDF form for download. Choose from 5-inner door upright freezers (Revco) or 4-inner door upright (TSX, Forma). Model numbers ending in A are 120V, 60Hz, D models are 208-230V, 60Hz and V models are 230V, 50Hz. Models with a WC at the end include water-cooled condensers. CAD and Revit format drawings will be available on this site in the near future; consult your sales representative for immediate needs for CAD and Revit.