Tube furnaces are typically used for processing small samples or heating in an inert atmosphere. The single and three-zoned constructions are designed to create precise temperature control.
Three-zone control enables the user to select a different temperature in each zone (e.g. for gas applications or material experiments). Some models offer split-hinge design, which easily allows you to change the tube.
Select the tube length and temperature range for your application
Temperature range | Product | Typical Application |
---|---|---|
Up to 1100°C | Lindberg Blue M Mini-mite | general lab use |
Up to 1200°C | Lindberg Blue M 1200°C split-hinge | industrial grade, 3 zone control available |
up to 1500°C | Lindberg Blue M 1500 °C heavy-duty box | general purpose |
up to 1700°C | Lindberg Blue M 1700 °C | industrial grade, research |
Temperature range | Product | Typical Application |
---|---|---|
up to 1100°C | Lindberg Blue M 1100 °C | industrial grade, 3 zone control available |
Up to 1200°C | Lindberg Blue M 1200°C split-hinge | industrial grade, 3 zone control available |
up to 1700°C | Lindberg Blue M 1700 °C box | industrial grade, research |
Temperature range | Product | Typical Application |
---|---|---|
up to 1100°C | Lindberg Blue M 1100 °C | industrial grade, 3 zone control available |
Up to 1200°C | Lindberg Blue M 1200°C split-hinge | industrial grade, 3 zone control available |
Resources
Heading
- Unity Lab Services
Access to our expert service team, information on support plans, parts and consumables resources, and other instrument services