Shake, rotate and mix it up
Thermo Scientific benchtop shakers, rotators, mixers, revolvers and rockers are robust enough for everyday use, yet provide precise sample agitation for all your applications. With a broad-range of digital speed and time settings that allow you to set your parameters and move on to your next task, you can be confident that your sample will shake, mix or pulse at the desired rate every time.
Benchtop shakers, rotators and mixers categories
Product Type | Description | Learn more | |
---|---|---|---|
Shakers | Microplate Shaker | ||
![]() | • Shaking of microplates for immunoassay, screening, magnetic bead, and other applications • Standard and enlarged platforms available — shake up to 6 microplates at a time. | Order | |
Teleshake Magnetic Shaking System | |||
![]() | • 100 to 2000rpm • Remote-controlled, wear-free magnetic shaking system • Suited for 96- and 384-well microtiter plates. | Order | |
Teleshake 1536-8 Magnetic Microtiter Plate Shakers | |||
![]() | • Up to 8500rpm • Wear-free magnetic drive • Remote controlled for integrated shaking applications • Suited for 1536- and 384-well microtiter plates. | Order | |
Accessories | Benchtop Shakers and Mixers | ||
![]() | • Assorted accessories | Order | |
Digital Microplate Shakers | |||
![]() | • Assorted accessories | Order |
Product Type | Description | Learn more | |
---|---|---|---|
Rotators | Tube Revolver / Rotator | ||
![]() | • Multiple rotating types with speed up to 40rpm • Adjustable paddle angle | ||
Digital Mini Rotators | |||
![]() | • Provides smooth, accurate and orbital mixing of various vessels in the labs | Order | |
Digital Waving Rotators | |||
![]() | • 360° orbital waving rotator for efficient sample mixing with accurate speed control and easy angle adjustment | Order | |
Cel-Gro Tissue Culture Rotator | |||
![]() | • Precisely mix test tube contents with rotating motion • Adjustable angle and variable speed supports mixing requirements for a diverse range of applications • Brushless, no maintenance motor provides smooth operation and worry free use | Order | |
Digital Mini Rotators | |||
![]() | • Assorted accessories | Order | |
Screws | Screws for Benchtop Shakers and Mixers | ||
![]() | • Assorted accessory screws | Order | |
Adaptors | General Power Adapters | ||
![]() | • For use with rockers, rotators, mixers | Order | |
Accessories | Accessories for Digital Rockers and Waving Rotators | ||
![]() | • Assorted rubberized accessories | Order |
Product Type | Description | Learn more | |
---|---|---|---|
Mixers | Basic Vortex Mixer | ||
![]() | • Provides fast, uniform mixing of samples with user-friendly mixing options • Continuous and touch modes • Stable for long-term operation | Order | |
Digital Vortex Mixer | |||
• Provides accurate speed control to continuously mix or pulse samples • Continuous and touch modes • Timer function for various applications • Stable for long-term operation | Order | ||
Thermal Mixer with Blocks | |||
![]() | • Heating and cooling capability to cover a wide range of applications | Order | |
Accessories | Thermal Mixer Blocks | ||
![]() | • A choice of blocks for microtubes and microplates. | Order | |
Power Adapters for Vortex Mixers | |||
![]() | • Power adapters which offer accurate and efficient sample mixing | Order | |
Accessories for Vortex Mixers | |||
![]() | • Assorted accessories | Order |
Product Type | Description | Learn more | |
---|---|---|---|
Rockers | Digital Rockers | ||
• Digital display with big platform, accurate speed control and quick angle adjustment | Order | ||
Digital Platform Rocker | |||
![]() | • Provides consistent rocking motion for mixing samples • Optional double-tier platform provides increased mixing capacity • Adjustable angle supports mixing requirements for a diverse range of applications | Order | |
Digital Tube Rocker | |||
![]() | • Provides smooth agitation and uniform specimen suspension for mixing samples in test tubes • Adjustable angle supports mixing requirements for a diverse range of applications • Optional double-tier platform provides increased mixing capacity | Order | |
Accessories | Accessories for Digital Rockers | ||
![]() | • Assorted rubberized accessories | Order | |
General Power Adapters | |||
![]() | • General power adapter offer accurate and efficient sample mixing | Order |
Product Type | Description | Learn more | |
---|---|---|---|
Revolvers | Tube Revolver / Rotator | ||
![]() | • Variable speed • Small footprint, flexible design and simple display help increase flexibility and effectiveness. | Order | |
Accessories | Accessories for Tube Revolver | ||
![]() | Accessories include variety of paddles | Order | |
Power Adapters for Tube Revolver / Rotator | |||
![]() | • Power adapters which offer accurate and efficient sample mixing | Order |
Product Type | Description | Learn more | |
---|---|---|---|
Rollers | Bottle/Tube Roller | ||
![]() | • Compact design for rolling multiple size tubes or bottles • Digital controlled speed ranging from 1- 80rpm • Safe to use in CO2 and/or humid environments • Maximize space by stacking two high with the optional stacking kit • Control without disturbing incubating samples with the optional remote control | ||
Accessories | Accessories for Bottle/Tube Roller | ||
![]() | • Accessories include O-rings, remote control and stacking kits | Order |
Product Type | Description | Learn more | |
---|---|---|---|
Shakers | Microplate Shaker | ||
![]() | • Shaking of microplates for immunoassay, screening, magnetic bead, and other applications • Standard and enlarged platforms available — shake up to 6 microplates at a time. | Order | |
Teleshake Magnetic Shaking System | |||
![]() | • 100 to 2000rpm • Remote-controlled, wear-free magnetic shaking system • Suited for 96- and 384-well microtiter plates. | Order | |
Teleshake 1536-8 Magnetic Microtiter Plate Shakers | |||
![]() | • Up to 8500rpm • Wear-free magnetic drive • Remote controlled for integrated shaking applications • Suited for 1536- and 384-well microtiter plates. | Order | |
Accessories | Benchtop Shakers and Mixers | ||
![]() | • Assorted accessories | Order | |
Digital Microplate Shakers | |||
![]() | • Assorted accessories | Order |
Product Type | Description | Learn more | |
---|---|---|---|
Rotators | Tube Revolver / Rotator | ||
![]() | • Multiple rotating types with speed up to 40rpm • Adjustable paddle angle | ||
Digital Mini Rotators | |||
![]() | • Provides smooth, accurate and orbital mixing of various vessels in the labs | Order | |
Digital Waving Rotators | |||
![]() | • 360° orbital waving rotator for efficient sample mixing with accurate speed control and easy angle adjustment | Order | |
Cel-Gro Tissue Culture Rotator | |||
![]() | • Precisely mix test tube contents with rotating motion • Adjustable angle and variable speed supports mixing requirements for a diverse range of applications • Brushless, no maintenance motor provides smooth operation and worry free use | Order | |
Digital Mini Rotators | |||
![]() | • Assorted accessories | Order | |
Screws | Screws for Benchtop Shakers and Mixers | ||
![]() | • Assorted accessory screws | Order | |
Adaptors | General Power Adapters | ||
![]() | • For use with rockers, rotators, mixers | Order | |
Accessories | Accessories for Digital Rockers and Waving Rotators | ||
![]() | • Assorted rubberized accessories | Order |
Product Type | Description | Learn more | |
---|---|---|---|
Mixers | Basic Vortex Mixer | ||
![]() | • Provides fast, uniform mixing of samples with user-friendly mixing options • Continuous and touch modes • Stable for long-term operation | Order | |
Digital Vortex Mixer | |||
• Provides accurate speed control to continuously mix or pulse samples • Continuous and touch modes • Timer function for various applications • Stable for long-term operation | Order | ||
Thermal Mixer with Blocks | |||
![]() | • Heating and cooling capability to cover a wide range of applications | Order | |
Accessories | Thermal Mixer Blocks | ||
![]() | • A choice of blocks for microtubes and microplates. | Order | |
Power Adapters for Vortex Mixers | |||
![]() | • Power adapters which offer accurate and efficient sample mixing | Order | |
Accessories for Vortex Mixers | |||
![]() | • Assorted accessories | Order |
Product Type | Description | Learn more | |
---|---|---|---|
Rockers | Digital Rockers | ||
• Digital display with big platform, accurate speed control and quick angle adjustment | Order | ||
Digital Platform Rocker | |||
![]() | • Provides consistent rocking motion for mixing samples • Optional double-tier platform provides increased mixing capacity • Adjustable angle supports mixing requirements for a diverse range of applications | Order | |
Digital Tube Rocker | |||
![]() | • Provides smooth agitation and uniform specimen suspension for mixing samples in test tubes • Adjustable angle supports mixing requirements for a diverse range of applications • Optional double-tier platform provides increased mixing capacity | Order | |
Accessories | Accessories for Digital Rockers | ||
![]() | • Assorted rubberized accessories | Order | |
General Power Adapters | |||
![]() | • General power adapter offer accurate and efficient sample mixing | Order |
Product Type | Description | Learn more | |
---|---|---|---|
Revolvers | Tube Revolver / Rotator | ||
![]() | • Variable speed • Small footprint, flexible design and simple display help increase flexibility and effectiveness. | Order | |
Accessories | Accessories for Tube Revolver | ||
![]() | Accessories include variety of paddles | Order | |
Power Adapters for Tube Revolver / Rotator | |||
![]() | • Power adapters which offer accurate and efficient sample mixing | Order |
Product Type | Description | Learn more | |
---|---|---|---|
Rollers | Bottle/Tube Roller | ||
![]() | • Compact design for rolling multiple size tubes or bottles • Digital controlled speed ranging from 1- 80rpm • Safe to use in CO2 and/or humid environments • Maximize space by stacking two high with the optional stacking kit • Control without disturbing incubating samples with the optional remote control | ||
Accessories | Accessories for Bottle/Tube Roller | ||
![]() | • Accessories include O-rings, remote control and stacking kits | Order |
Watch to learn about Thermo Scientific shakers, rotators and mixers
Resources
- Brochure: Thermo Scientific Benchtop Shakers [EN], [JP]
- Digital Mini Rotator User Manual [EN], [DE], [ES], [FR], [IT]
- Digital Rocker User Manual [EN], [DE], [ES], [FR], [IT]
- Digital Vortex Mixer User Manual [EN], [DE], [ES], [FR], [IT]
- Digital Microplate Shaker User Manual [EN], [DE], [ES], [FR], [IT]
- Digital Waving Rotator User Manual [EN], [DE], [ES], [FR], [IT]