Search Thermo Fisher Scientific
Count on IntelliStack Incubator Shakers and CO2Incubator Shakers to help provide accurate, adjustable and reproducible growth conditions for your mammalian, bacterial and yeast cell culture and other applications. Whether you need one unit or multiple, you can customize your setup according to your workflow requirements and processing volume as your needs evolve. See for yourself how this fully integrated shaking solution with touchscreen environmental control features stacks up.
IntelliStack Incubator Shakers and CO2 Incubator Shakers can help streamline your workflow and provide peace of mind that your results are reliable and reproducible.
Ease of use: The 10" color touchscreen interface is intuitive and user-friendly, allowing for easy control and adjustment of parameters such as temperature, speed, CO2 and humidity. All parameters can be viewed, set, started, and stopped from the main screen.
Real-time monitoring: The smart system shares information in real time, allowing users to see both the set value and current value for each parameter. This enables better monitoring and control of the incubation process to help ensure optimal conditions for experimentation and sample growth.
Programmability: From the home screen, you can create and program customized protocols with up to 12 steps. This flexibility allows for precise control over incubation time, temperature, speed and other parameters, helping ensure reproducibility and accuracy in your processes. You can also operate in a continuous timer mode by choosing manual mode.
Data management: Users can easily retrieve run data, alarm history and operation logs in PDF or CSV format.
User access control: With this feature, you can assign different levels of access for authorized personnel to enhance security and enable only authorized individuals to make changes to critical parameters.
IntelliStack Incubator Shaker
When your cell culture and microbial growth depend on consistent and reliable results, count on the IntelliStack Incubator Shaker. It allows you to control and maintain the internal temperature and humidity, enabling the right environmental conditions for your samples to thrive.
IntelliStack CO2 Gassing Incubator Shaker
When your experimental progress depends on optimal cell growth, gas exchange and reproducibility, count on the IntelliStack CO2 Gassing Incubator Shaker to fulfill your workflow needs.
Each IntelliStack unit features a large loading platform and interior chamber. A stack of three units can fit up to 96 of the 500 ml flasks, 45 of the 2 L flasks or 18 of the 5 L flasks. All IntelliStack varieties can be stacked.
Orbit (mm) |
Voltage |
Swing door direction |
Catalog No. |
---|---|---|---|
25 |
120V, 50/60Hz |
Down swing door |
|
25 |
120V, 50/60Hz |
Up swing door |
|
25 |
220V, 50/60Hz |
Down swing door |
|
25 |
220V, 50/60Hz |
Up swing door |
|
50 |
120V, 50/60Hz |
Down swing door |
|
50 |
120V, 50/60Hz |
Up swing door |
|
50 |
220V, 50/60Hz |
Down swing door |
|
50 |
220V, 50/60Hz |
Up swing door |
Orbit (mm) |
Voltage |
Swing door direction |
Catalog No. |
---|---|---|---|
25 |
120V, 50/60Hz |
Down swing door |
|
25 |
120V, 50/60Hz |
Up swing door |
|
25 |
220V, 50/60Hz |
Down swing door |
|
25 |
220V, 50/60Hz |
Up swing door |
|
50 |
120V, 50/60Hz |
Down swing door |
|
50 |
120V, 50/60Hz |
Up swing door |
|
50 |
220V, 50/60Hz |
Down swing door |
|
50 |
220V, 50/60Hz |
Up swing door |
For Laboratory Use. It is the customer’s responsibility to ensure that the performance of the product is suitable for customers’ specific uses or applications.