reciprocating shakers
Facilitate repeatable results with chemical extractions, sample mixing and silver staining of polyacrylamide gels in addition to bacterial and yeast cultures with reciprocating and dual action shakers.
Choose shaker    Choose accessories
Reliability
reliable reciprocating shakers
We stand behind our shakers with an industry leading warranty that provides 2-year coverage on labor, 5 years on parts, and a full 10-year warranty on the drive mechanism.
Simplicity
easy-to-use reciprocating shakers
Units feature an easy-to-use analog operating system and variable speed control for gentle-to-vigorous agitation.
Flexibility
flexible reciprocating shakers
An added reciprocating function, analog operating system and choice of three platforms make this an affordable laboratory shaker with increased capacity for a wide range of applications.
Choose a reciprocating shaker
  MaxQ 2506 Reciprocating Shaker MaxQ 2508 Dual-Action Orbital Reciprocating Shaker
  maxq 2506 shaker maxq 2508 shaker
  Provides back-and-forth motion or circular orbital motion for a variety of applications. On dual action models, a control lever changes shaking motion while the unit is stationary or in operation.
  Learn more Learn more
Maximum vessel size 6L Erlenmeyer flask
6L Erlenmeyer flask
Available universal platforms 11 x 13 in. 11 x 13 in.
Speed 15 to 500 ±1rpm 40 to 400rpm
Exterior dimensions
(H x W x D)
7.2 x 13.5 x 17.4 in.
(18.3 x 34.3 x 44.2cm)
7 x 13.5 x 17.4 in. (17.8 x 34.3 x 44.2cm)
Orbit Adjustable 0.5 to 1.5 in. (1.2 to 3.8cm) 0.4 in. (1.2cm) circular or fixed stroke angle
Weight capacity 15 lb. (6.8kg) 15 lb. (6.8kg)
Voltage 120V 120V
Accessories Platforms ›
Clamps ›
Racks & Misc ›
Platforms ›
Clamps ›
Racks & Misc ›
Top
Want some help making a decision? Talk with one of our expert sales representatives.     Contact us
Optimize your reciprocating shaker
reciprocating shaker platforms
Universal platforms maximize flexibility by mixing different size labware on a single platform. Go ›
reciprocating shaker clamps
Universal clamps maximize efficiency and productivity by mixing and matching vessel types with your universal platforms and customizing your shaker to fit your unique needs. Go ›
reciprocating shaker racks
Test tube racks and accessories outfit your shaker with holders that adjust, stack, and interlock to accommodate a wide variety of applications and objectives, and to keep conditions clean and safe. Go ›
Universal platforms
Choose a platform that best fits your needs with flexibility for using a variety of labware on a single platform. Purchase clamps separately. Pick your ideal dimensions (L x W) below.
Universal clamps
Mix and match to customize your universal platform for your unique needs. Clamps sold as individual units.
Test tube racks & accessories
Top
The clamp alternative
lab shaker flask mat
Reliable, easy-to-use "Sticky Mats"
Thermo Scientific Adhesive Flask Mats, or "sticky mats," provide a flexible, convenient alternative for attaching a variety of vessels to any orbital shaker.
Top 3 best practices for using “sticky mats” on an orbital shaker
Order now
Top
Related products
orbital shaker
co2 resistant shaker
reusable lab plasticware
co2 incubators