Download the waterbook
Broad range of stills
Effectively remove most inorganic solids, all organics with a boiling point greater than water (100°C), all bacteria, and pyrogens. Ranging in size from 1.4 to 38L/hr and available in tin or glass, our water still models offer fully automatic options or pretreatment options to minimize scaling.
View the Waterbook, browse our complete portfolio or let one of our experts help you make a selection.
Choosing the right distillation for your lab application
Choosing the right distillation for your lab application
Of the broad array of water purification technologies available, it is important that users select the most appropriate method for their applications.
Download brochure 
Lab Water System Selector Tool	
Support
Unity Lab Services:  Access to our expert service team, information on support plans, parts and consumables resources, and other instrument services.