Large Central Water Purification Systems
Customized solutions for large pure water volume requirements
Ensure consistent results for your most critical and everyday applications. Thermo Scientific large central water purification systems are designed to produce reliable pure water meeting ISO 285, ASTM Type 2, Clinical Laboratory Reagent Water (CLRW) and European Pharmacopeia standards.
Central systems can replace numerous individual systems, while reducing costs by eliminating the need for ion exchanger regeneration. They meet daily volume requirements from 300 to > 100,000 L/day and support a number of applications.
Featured water purification systems categories
Our ready-to-connect reverse osmosis systems for the demineralization of softened or hardness-stabilized tap water meet pure water volume requirements up to 25,000 L/day.
When conductivity below 1 μS/cm is required, select a large reverse osmosis water purification system with downstream electrodeionization (EDI) technology.
Maintain your large central water purification system with accessories and consumables, including carbon filters, cartridges, and softening systems.
Unity Lab Services: Access to our expert service team, information on support plans, parts and consumables resources, and other instrument services.