Search
Thermo Scientific Forma Vertical Light Refrigerated Reach-In Chambers offer a wide temperature range, outstanding temperature stability and uniformity, and versatile lighting modules. These units are ideal for applications requiring precise temperature and lighting control, without the need for humidity control.
Temperature
Temperate ranges of 0°C to 60°C with light, 10°C to 50°C without light, setting in 0.1°C steps.
Versatile lighting
Lighting modules for stability testing according to ICH part Q1B guidelines. Lighting modules for plant growth emit more red/blue frequency and those tailored to insect growth simulate daylight, dawn, and dusk cycles.
Programmable
Simple, easy to control programming are features across models.
|
Vertical Light Refrigerated Reach-In Chamber |
Size |
29 cu. ft. |
Exterior (W x D x H) |
965 x 2248 x 1308 mm 38 x 32 x 88.5 in. |
Interior (W x D x H) |
787 x 686 x 1524 mm / 31 x 27 x 60 in. |
Net weight |
318kg / 700 lbs |
Shipping weight |
386kg / 850 lbs |
Set Light and Temperature |
|
Max # of light modules |
3 |
Temperature range |
0°C to 60°C with light, 10°C to 50°C without light |
Controller, sensor |
Watlow Microprocessor - PID, with RTD sensor |
Display |
Digital dual LED, 0.1°C steps readability & setability |
Temperature Uniformity |
≤±0.3°C with light off ≤±3.0°C - with light on, @ 20°C to 37°C @ 75% RH with 2 light modules |
Temperature Stability |
≤±0.1°C with light off ≤±0.5°C - with light on, @ 20°C to 37°C @ 75% RH with 2 light modules |
Electrical |
|
Voltage |
120V, 60Hz, 16.0 A, 3 Wire (incl. Ground) single phase |
Plug |
NEMA 5-20P |
Power switch |
2-Pole |
Interior Convenience receptacles |
Optional |
Alarm Contacts |
Temp. deviation and power failure; Common, NO, NC |
Available Options |
Shelving Systems Access Ports
|
Order Number |
|