Heratherm Microbiological Incubator
Your samples safe inside
Your biological samples are often irreplaceable. That makes your microbiological incubator critical to your success. Your work is challenging enough. Surround your life with some certainty with Thermo Scientific™ Heratherm™ microbiological incubators—available in four models, providing three incubator airflow technologies in a choice of six sizes.   
Download brochure ›
Heratherm Refrigerated Incubators
Introducing Heratherm refrigerated incubators. With no harmful refrigerants and the use of Peltier technology, these units are energy efficient  while maintaining the precise temperatures you need. Learn about the benefits of energy savings in your lab for refrigerated incubation.
Learn More ›
Heratherm microbiological incubator categories
Thermo Scientific Heratherm general protocol incubators are designed for routine applications in pharmaceutical, medical, food, and research laboratories. They come in three sizes, offering a flexible shelf system and easy-to-use interface.
Thermo Scientific Heratherm advanced protocol incubators provide exceptional temperature performance for demanding applications. Their innovative design includes dual convection and an advanced digital timer with weekly, real-time, and hourly settings.
Thermo Scientific Heratherm advanced protocol security incubators incorporate additional safety features to protect your samples, including a lockable incubator door for restricted access and an audible alarm if the door is left open. An automatic decontamination cycle helps to ensure the unit is clean after every experiment.
Thermo Scientific Heratherm compact incubators are the most compact unit of the Heratherm microbiological incubator family. They feature an 18 L capacity—ideal for a personalized workspace—as well as high-temperature accuracy and an internal light source.
Featured video
Heratherm microbiological incubators
Watch this video to learn how our Heratherm microbiological incubators can meet your application needs.