Reliable and affordable fluorescence and luminescence plate readers
The Thermo Scientific Fluoroskan Microplate Fluorometer is a compact, robust fluorescence plate reader built to run variety of fluorometric and FRET assays with excellent performance. The Thermo Scientific Luminoskan Microplate Luminometer is a sensitive, DLReady certified luminescence plate reader built to perform superbly for a variety of luminescence applications, including flash, glow and BRET assays. The DLReady-certified Thermo Scientific Fluoroskan FL Microplate Fluorometer and Luminometer is a dual-mode versatile workhorse for fluorometric and luminometric assays. Fluoroskan, Fluoroskan FL and Luminoskan instruments are all equipped with powerful Thermo Scientific SkanIt Software, offering increased usability for setting up protocols and data analysis. Use validated protocols from the growing SkanIt Cloud Library to help save time and make your workflow more efficient.
Features
Reagent dispensers: simultaneous dispense and read
Reagent dispensers enable easy and accurate reagent addition simultaneously as the instrument reads—a critical feature for flash-type reactions. All Fluoroskan and Luminoskan models are offered with three options of associated reagent dispensers: two dispensers, one dispenser, or no dispensers.
Flexibility to read 6- to 384-well plates
Fluoroskan, Fluoroskan FL and Luminoskan plate readers allow you to choose your sample format depending on your unique research and throughput needs.
High sensitivity for both top and bottom reading
Fiberless direct illumination optics for both top and bottom reading. Top reading is great for homogeneous, BRET and FRET assays, whereas bottom-reading becomes very important for non-homogeneous and cell-based assays. Bottom reading shortens the distance between the cell layer at the bottom of the well and the detector and minimizes any effect from the cell culture medium.
Dynamic range >6 decades
Reliably measure high and low signal samples with Fluoroskan and Luminoskan readers that provide both sensitivity and broad dynamic range in the same run.
Multipoint well reading
Read multiple locations in a single well by stipulating in SkanIt Software where in the well you want to read. This is especially important for non-homogeneous mixtures and cell-based assays where cells are not uniformly distributed in each microplate well. Multipoint measurements on Thermo Scientific fluorescence plate readers can also be used as an alternative or quantitative complement to imaging fluorospot assays.
Incubation and shaking
Incubate up to 45°C with orbital shaking.
Auto-calibration and self-diagnostics
Enables accurate fluorescence data with every run. If something isn't quite right, all three reliable fluorescence and luminescence plate readers can tell you before your run.
Fluorescence plate readers
Dedicated Fluoroskan microplate fluorometer and dual-mode Fluoroskan FL fluorometer and luminometer
The high-performance fluorometers in Fluoroskan and Fluoroskan FL microplate readers make them ideal fluorescence plate readers for life science research applications such as:
- fluorometric protein and enzyme studies
- molecular interactions
- nucleic acid quantification
- reporter gene assays
- fluorometric kinase studies
- immune- and cell-based assays
Luminescence plate readers
Dedicated Luminoskan microplate luminometer and dual-mode Fluoroskan FL fluorometer and luminometer
The quality engineered luminometers in Luminoskan and Fluoroskan FL readers make them high-performing luminescence plate readers for life science research applications such as:
- cell viability studies
- cytotoxicity studies
- ATP determination assays
- luciferase assays
- reporter gene assays
- GPCR/calcium activation
SkanIt software
Fluoroskan, Fluoroskan FL and Luminoskan plate readers are all controlled by intuitive and powerful Thermo Scientific SkanIt Software. SkanIt Software supports optimal use of instrument features with a visual workflow, intuitive virtual pipetting tool, effortless data analysis and exporting capabilities. SkanIt software is a powerful multi-instrument microplate reader software that provides excellent usability and flexibility even for the most challenging assays.
Another bonus, SkanIt Software is used with all Thermo Scientific readers, making it easy for labs to have multiple readers run by the same easy-to-use program on a single computer. In addition, you can purchase a SkanIt upgrade package to run the latest version of SkanIt Software on certain older Thermo Scientific Fluoroskan, Fluoroskan FL and Luminoskan models.
Figure 1. The session tree is the main area for navigation within the software. The measurement protocol and the results calculations are defined within a user-friendly hierarchy.
Figure 2. The innovative Virtual Pipetting Tool makes it easy to define samples in the plate layout.
SkanIt cloud library
This growing library of validated, pre-built protocols makes it easy to run assays on Thermo Scientific plate readers. Among the list of protocols available are various fluorescence and luminescence assays for measurement of:
- cellular viability, proliferation and cytotoxicity
- ATP determination and luciferase activity
- intracellular calcium, magnesium and pH
- luminescence monitoring transfection efficiency and detecting expression of recombinant genes
- BRET energy for specific protein pair interactions
- Ca2+ flux assays
- cell proliferation
- cytotoxicity
- multidrug resistance
- cell adhesion
- DNA quantitation
- reporter gene assays
- hybridization assays
- ion channel assays
- quantitation of PCR products
- FRET assays
- molecular beacon assays
- immunoassays
- enzyme activity
- neonatal screening
- bacterial quantification
- phagocytosis
- oligonucleotide assays
- ADMEtox
- fluorometric protein and enzyme studies
- molecular interactions
- nucleic acid quantification
- reporter gene assays
- fluorometric kinase studies
- immune- and cell-based assays
- drug discovery
- flash luminescence assays
- glow luminescence assays
- BRET and nanoBRET assays
- luminescence enzyme assays
- luminescence immunoassays
- cell viability studies
- cell proliferation studies
- cytotoxicity studies
- ATP determination assays
- microbial contamination assays
- luciferase assays
- reporter gene assays
- GPCR/calcium activation assays
- drug discovery
Highly sensitive DNA quantification
Fluoroskan Microplate Fluorometer, when used with the Invitrogen Quant-iT dsDNA high sensitivity assays, offers excellent sensitivity even in the lower DNA concentration range in the microplate format. This fluorescence plate reader detects as little as 0.2 ng of λ DNA (Figure 3), corroborating kit specifications.
Figure 3. Highly sensitive DNA quantification. Fluorescence measurements were taken of a standard dilution of λ DNA, labeled using the Quant-iT dsDNA Assay Kit, high sensitivity distributed in Nunc F96 MicroWell Black Polystyrene Plates and quantified using the Fluoroskan Microplate Fluorometer and SkanIt Software. Calculated limit of detection (LOD), as determined by the IUPAC standard 3*SD method, was 0.04 ng in this experiment.
Application note: Sensitive fluorometric quantification of DNA in microplate format
Download the application note and learn about sensitivity differences between UV-Vis and Fluorescence detection of nucleic acids, Quant-iT kits, and how Thermo Scientific Fluoroskan and Fluoroskan FL fluorescence plate readers perform sensitive fluorometric quantification of DNA in a microplate format.
Pharmacological readout is consistent with both Fluoroskan FL and Varioskan LUX plate readers
Staurosporine is a global kinase inhibitor; niclosamide induces apoptosis through a mitochondrial intrinsic pathway; and gambogic acid causes apoptosis induction by inhibiting proteasome function leading to apoptosis and cell death. In a standard dose-response experiment involving these three drugs, the pharmacological readout is the EC50, effective concentration 50, which is the concentration of the drug that gives half the maximal response. The maximum response of these drugs is complete cell death, so EC50 is the drug concentration at which half of the cells remained viable, as measured by Invitrogen alamarBlue Cell Viability Reagent. Regardless of which of our plate readers you use—the filter-based Fluoroskan or monochromator-based Varioskan LUX—the data agree (Figure 4).
Figure 4. Dose-response curves for cultured A549 monolayers treated with three model drugs: staurosporine, niclosamide and gambogic acid. A549 cells were plated in a 2D monolayer on Nunc cell culture plates, treated with varying concentrations of each drug for 24 hours. Viability was assayed using alamarBlue reagent and measured on two Thermo Scientific plate readers: the Varioskan LUX Multimode Microplate Reader and the Fluoroskan FL Microplate Fluorometer and Luminometer. These data demonstrate that the pharmacological readout is the same regardless of which reader is used.
All three drugs have a different EC50, but the two different microplate readers show the same EC50 for each drug. This data validates each reader’s accuracy in measuring and calculating pharmacological data (Table 1).
EC50 (µM) | ||
---|---|---|
Varioskan LUX | Fluoroskan FL | |
Staurosporine | 6.786 | 6.639 |
Gambogic acid | 0.7222 | 0.7217 |
Niclosamide | 0.5898 | 0.5705 |
Flash-type ATP reactions with kinetic measurements
Get powerful luminescence detection of a flash-type ATP reaction performed by two of our reader models: the dedicated Luminoskan luminescence reader and the luminescence module of the dual-mode Fluoroskan FL microplate reader. Without access to the right equipment, researchers often rely on less-sensitive glow-type assays to detect ATP reactions. With simultaneous dispense-read technology, Luminoskan and Fluoroskan FL readers offer increased sensitivity with sub-attomole detection limits (Figure 5).
Figure 5. ATP standard curves generated with a flash-type luminescence ATP assay. ATP standard curves with the same slope generated with Luminoskan Luminescence Plate Reader and Fluoroskan FL Fluorescence and Luminescence Plate Reader exhibit detection success across a broad ATP concentration range as well as <1 amol detection limits.
Application note: Ultrafast flash kinetic assays with Luminoskan and Fluoroskan FL microplate readers
Download the technical application note and learn about the differences between flash-type and glow-type luminescence assays, protocols, and how Thermo Scientific Luminoskan and Fluoroskan FL luminescence plate readers perform accurate, timely kinetic measurements for rapid, flash-type reactions.
Technical specifications for the Fluoroskan, Luminoskan, and Fluoroskan FL microplate readers
Fluoroskan | Luminoskan | Fluoroskan FL | |
Fluorometry | |||
---|---|---|---|
Excitation wavelength range | 320–700 nm | 320–700 nm | |
Emission wavelength range | 360–800 nm | 360–670 nm | |
Excitation filters | Up to 8 filters in the excitation filter wheel; 355 nm and 485 nm filters included as standard. Other filters available upon request. | Up to 8 filters in the excitation filter wheel; 355 nm and 485 nm filters included as standard. Other filters available upon request. | |
Emission filters | Up to 8 filters in the emission filter wheel; 460 nm and 538 nm filters included as standard. Other filters available upon request. | Up to 6 filters in the emission filter wheel. 460 nm and 538 nm filters included as standard. Other filters available upon request. | |
Sensitivity | 2 fmol fluorescein/well in a black 96-well plate | 2 fmol fluorescein/well in a black 96-well plate | |
Dynamic range | >6 decades | >6 decades | |
Luminometry | |||
Spectral range | 270–670 nm | 270–670 nm | |
Sensitivity | 10 amol ATP/well using flash reaction, white 384-well plate | 40 amol ATP/well using flash reaction, white 384-well plate | |
Dynamic range | >6 decades | >6 decades | |
Luminometric filters | Up to 6 luminometric filters in the filter wheel. All filters available upon request. | ||
Dispensing | |||
Number of dispensers | Up to 3* | Up to 3* | Up to 3* |
Dispensing volume | 4–995 μL in 1 μL increments | 4–995 μL in 1 μL increments | 4–995 μL in 1 μL increments |
Dispensing speed | 30 s, 96-well plate, 5 μl/well | 30 s, 96-well plate, 5 μl/well | 30 s, 96-well plate, 5 μl/well |
General features | |||
Plate types | 6- to 384-well plates | 6- to 384-well plates | 6- to 384-well plates |
Measurement speed | 18 s, 96-well plate 60 s, 384-well plate |
18 s, 96-well plate 60 s, 384-well plate |
18 s, 96-well plate 60 s, 384-well plate |
Wavelength selection | Filters | Filters | Filters |
Light source | Quartz-halogen lamp | Quartz-halogen lamp | |
Detector | Photomultiplier tube | Photomultiplier tube | Photomultiplier tube |
Incubator | From ambient + 3°C to 45°C, at ambient 25°C | From ambient + 3°C to 45°C, at ambient 25°C | From ambient + 3°C to 45°C, at ambient 25°C |
Plate shaking | Orbital | Orbital | Orbital |
User interface | PC software (SkanIt software) | PC software (SkanIt software) | PC software (SkanIt software) |
Computer interface | USB 2.0 | USB 2.0 | USB 2.0 |
Dimensions (H x W x D) | 340 x 420 x 420 mm 13.4 x 16.5 x 16.5 in. |
340 x 420 x 420 mm 13.4 x 16.5 x 16.5 in. |
340 x 420 x 420 mm 13.4 x 16.5 x 16.5 in. |
Weight | Basic unit 21 kg (46 lb) 2 optional dispensers add 2.4 kg |
Basic unit 20.5 kg (45 lb) 2 optional dispensers add 2.4 kg |
Basic unit 21 kg (46 lb) 2 optional dispensers add 2.4 kg |
* Third dispenser available with service installation.
Ordering information
Resources
Services & support
Technical support
Starting tips and troubleshooting, visit our Support Centers:
Service plans
We provide a full range of service options for all Microplate Readers. Visit our Service Selection Tool today to receive a customized quote.
Educational resources
Application notes, handbooks, webinars, white papers, how-to videos, case studies, scientific posters, product selection guides, visit our Learning Centers:
Microplate Reader Software