SkanIt software screenshot
The all-in-one plate reader control and analysis software
Powerful, intuitive Thermo Scientific SkanIt Software is a user-friendly, productivity-boosting, error-preventing platform that drives Thermo Scientific microplate readers. Two editions of the reader control and analysis software are available: SkanIt Software Research Edition is free with purchase of a Thermo Scientific plate reader, and the optional SkanIt Software Drug Discovery Edition that enables compliance with 21CFR Part 11 can be purchased with your order or separately.
Download SkanIt Software
SkanIt Software makes it easy to have just one or multiple Thermo Scientific microplate readers in your lab. You can choose to run all your instruments with SkanIt Software from a single computer or multiple computers, and since the software comes with unlimited installations, you can use any computer you like to perform data analysis from wherever you like.
Currently available plate reader models include the Thermo Scientific Varioskan LUX Multimode Microplate Reader, the Thermo Scientific Fluoroskan FL Microplate Fluorometer and Luminometer, the Thermo Scientific Fluoroskan Microplate Fluorometer, the Thermo Scientific Luminoskan Microplate Luminometer, the Thermo Scientific Multiskan SkyHigh Microplate Spectrophotometer, and the Thermo Scientific Multiskan FC Microplate Photometer. Some of the predecessor models of Thermo Scientific Fluoroskan Ascent FL, Fluoroskan Ascent, and Luminoskan Ascent may also be upgraded.
Key benefits of SkanIt Software
Easy-to-use plate reader software
Logical software structure
  • The logical structure of the microplate reader software makes it easy to use
    • Intuitive Virtual Pipetting Tool (see figure)
    • Session tree (see figure)
  • No need for training to use the instrument
    • The only instruction we needed to give was the computer password
    • The software is self-explanatory
  • Easy and straightforward to create a new protocol adapted to different experimental conditions
Merge data
The ability to merge data is helpful in a variety of assay types and improves data visualization on graphs. This feature is especially useful with Fluoroskan and Luminoskan instruments when kinetic assays with dispensing are performed. You may find this feature particularly valuable if you:
  • Read baseline kinetically before reagent addition, dispense reagent to activate the reaction, and continue reading the kinetic signal.
  • Conduct assays with multiple reagent additions.
  • Perform calcium flux measurements (Fluo3, Fluo4, Fura2).
Easy file access
  • Double clicking from file icon opens software just like a Microsoft Office file.
  • Intuitive search from “Open” within SkanIt Software is friendly for handling files.
  • Save anywhere you want—desktop, network drive, USB, cloud-based storage, etc.—just like a Microsoft Office file.
Flexible use in and out of the lab
  • Multi-instrument software platform compatible with all Thermo Scientific microplate readers
    See microplate reader selection
  • Robotic automation interface
    See lab automation solutions
  • Royalty-free license software for unlimited installation to multiple computers
    • No recurring cost to use software
    • Less hassle, with fewer restrictions
    • Analyze data from anywhere
    • Helpful in a multi-user environment
SkanIt Software Instruments
SkanIt microplate reader software helps prevent errors
  • Prompt to rename experiments prevents mix-up/loss of data
  • Measurement data within executed session is never lost, unless the file is intentionally deleted by the user. This means you can simply try different steps and “play” within the software as you design your experiments.
  • Drug Discovery Edition with features for compliance with FDA 21 CFR Part 11
  • Back-calculation of standards for verification of results (sanity check)
Plate reader software that helps increase productivity
  • High processing speed
  • Ability to analyze your data on a different computer than the main lab computer with royalty-free license software
  • Easy access to data: save a file wherever you want—desktop, network drive, USB, cloud-based storage, etc.—and simply double-click to open SkanIt files just like you would for Microsoft Office files
  • Extensive analysis tools for many common workflows
  • Automatic data loading into custom-defined Microsoft Excel template
Distinctive SkanIt Software features
What is SkanIt Cloud Library?
SkanIt Cloud Library is a collection of validated sessions—protocols that have analysis steps with real data and calculations—within Thermo Scientific SkanIt Software, which controls all Thermo Scientific microplate readers. Users can access this SkanIt Cloud Library for performing immediate measurements or to use the sessions as templates to create their own assay protocols. SkanIt Cloud Library is also a data source for training (including self-training) of new users of SkanIt Software. The library contains protocols and representative examples of the most frequently used optical detection technologies: absorbance, fluorescence, time-resolved-fluorescence, luminescence and AlphaScreen. Each SkanIt Cloud Library session has complete sets of measurements with the required calculations and graph processing steps from real assays. Instructions for running the assays are also provided. The library currently contains hundreds of protocols and continues to expand as new applications emerge.
How to use SkanIt Cloud Library?
SkanIt Cloud Library can always be accessed with the latest SkanIt version(s) when the PC is connected to Internet. The software is royalty free, so users of compatible Thermo Scientific microplate readers can freely install SkanIt Software on multiple PCs. Once SkanIt Software is installed, SkanIt Cloud Library is readily available for use. The Cloud Library icon is visible in the Home menu of the software (a), as well as in the Main menu (b), as shown below:
SkanIt microplate reader software home menu with red box around Cloud Library icon
(a)
SkanIt microplate reader software main menu with red box indicating Cloud Library icon
(b)
With the newest version of SkanIt Software, SkanIt Cloud Library has become available in a redesigned format, making it simpler to search protocols by instrument, detection technology, plate format, or other relevant search terms (if you click to enlarge the image of the search tool, you see an example in which "Invitrogen" was used as a search term). After selecting the Cloud Library, the entire list of protocols appears and the specific term of interest can be typed in the search bar. The most common search terms are already listed just below the search bar—simply click for a faster search. Additional search terms, or tags, can be easily added by clicking the [ + ] button next to the search bar to quickly customize future searches. Popular Thermo Fisher Scientific kits such as Invitrogen CyQuantInvitrogen PrestoBlue Cell Viability Reagent, or Invitrogen Quant-iT dsDNA High-Sensitivity (HS) Assay Kit can also be easily located. Some CyQuant sessions contain data with 3D spheroids.
Once the selected cloud session is opened, general technical guidance for how to perform the assay can be found in the “Notes” section. Instructions on the “Plate Layout,” “Protocol,” and “Results” sections are also included.
There are a couple of different ways to save your session, depending on your needs. If you do not need to change the protocol steps, click “Save” or “Save As” to keep the measurement data that was included in the Cloud Library session. Clicking “Save” or “Save As” makes the content of the session (except the protocol steps) editable as needed. If you need to change the protocol steps, click “Save As New” to enable that flexibility. When you click “Save As New,” the existing results data will be removed and all the content of the session will be editable as needed, including the protocol steps.
What are the benefits of using the Cloud Library?
SkanIt Cloud Library facilitates setting up assays using Thermo Scientific microplate readers and:
  • Minimizes the time needed to successfully conduct measurements, helping boost productivity.
    Learn more ways SkanIt Software helps improve productivity
  • Makes it easier to design experiments and learn how to run new protocols with technical descriptions of the assays, offering general guidance and assistance with assay performance and data processing.
    Learn more ways SkanIt Software is easy to use
  • Provides access to examples of the most frequently used microplate assays in life sciences, such as: quantification of nucleic acids, quantification of proteins, cell health (cytotoxicity, proliferation), cell signaling, reporter gene and enzyme assays, as well as ion indicators.
  • Features fully customizable protocol templates that can be customized to specified plate layouts, desired measurement, dispensing, shaking, or incubation steps, and any data processing and visualization steps needed to answer important research questions.
  • Is easily searchable via a search engine where sessions can be found quickly by using pre-defined tags or other relevant search keywords.
  • Contains hundreds of validated sessions—comprised of real assay data generated by our scientists—and the Cloud Library continues to expand as new assays and applications emerge.
Learn more about SkanIt Software
Save resources with FluoroSpot pre-screening feature of SkanIt software for microplate readers
For the first time ever, a microplate reader is able to give you an indication of the success of your FluoroSpot assay before you spend resources imaging the plate. This is a new technology offered by Thermo Scientific fluorescence microplate readers and made possible by the multipoint top-reading capability introducted with the 6th edition of SkanIt Software.
Often performed in cancer, infectious disease, and immunology research, a remarkable feature of the ELISpot assay is its sensitivity. High sensitivity is important in these fields because, for example, antigen-specific T cells are typically present at low frequencies in vivo. With ELISpot, it is possible to detect a single cytokine-producing cell from as many as one million bystander cells.
Detection of cell-secreted proteins can also be achieved using fluorescence-labeled detection reagents (including streptavidin fluorescent conjugates), and in this case the assay is known as FluoroSpot. ELISpot and FluoroSpot assays use the same protocol shown below for coating plates with capture antibodies, cell culture, and stimulation and washing steps. FluoroSpot has an advantage over ELISpot in that it is more suited for the simultaneous detection of multiple analytes, also known as multiplexing.
The ELISpot/FluoroSpot assay workflow shown graphically followed by example images of ELISpot and FluoroSpot wells
The ELISpot/FluoroSpot assay workflow.
FluoroSpot pre-screening with Thermo Scientific fluorescence microplate readers
FluoroSpot pre-screening with Thermo Scientific fluorescence microplate readers
Learn how checking your FluoroSpot plates with the Thermo Scientific exclusive pre-screening feature enables you to identify low- and high-secreting samples quickly and reduce subjectivity prior to sending for imaging.
Read the app note
Fluorescence SpectraViewer
Set up your fluorescence assays quickly and easily using the Fluorescence SpectraViewer tool.
  • The Fluorescence SpectraViewer tool—integrated in SkanIt Software—is based on the well-known and popular Fluorescence SpectraViewer on our website.
  • This dye library inside SkanIt software contains the fluorescence spectra of about 500 commonly used dyes and their excitation and emission peak wavelengths.
  • Simply choose the dye used in your assay and select the excitation and emission wavelengths for your SkanIt session, based on SpectraViewer data.
Screenshot of Fluorescence SpectraViewer tool showing the settings and excitation and emission peaks for calcium bound Fluo-4
More key features of SkanIt microplate reader software
Easy file-based user experience
  • Simply double-click SkanIt files to open them in SkanIt software—a similar experience to opening any Microsoft Office file.
  • No need for complicated import/export of SkanIt files using the latest edition of SkanIt Software.
Even faster data processing
  • Some backend software enhancements have given way to even faster performance, helping quicken your data acquisition and analysis.
PLA software interface with Stegmann Systems GmbH
  • Stegmann Systems PLA software is the standard reference for PLA calculations in the world.
  • Data is exported from SkanIt in a single click as an Excel file and imported into PLA 3.0 using Stegmann’s Microsoft Excel Data Aquisition Package. Then PLA calculations are performed using Stegmann PLA 3.0 software.
Stegmann-systems-pla-with-thermo-scientific-microplate-reader-data-from-skanit-software
Stegmann Systems PLA 3.0 software analyzing neuramidase inhibition assay data exported from SkanIt software for Thermo Scientific microplate readers.
What is SkanIt Cloud Library?
SkanIt Cloud Library is a collection of validated sessions—protocols that have analysis steps with real data and calculations—within Thermo Scientific SkanIt Software, which controls all Thermo Scientific microplate readers. Users can access this SkanIt Cloud Library for performing immediate measurements or to use the sessions as templates to create their own assay protocols. SkanIt Cloud Library is also a data source for training (including self-training) of new users of SkanIt Software. The library contains protocols and representative examples of the most frequently used optical detection technologies: absorbance, fluorescence, time-resolved-fluorescence, luminescence and AlphaScreen. Each SkanIt Cloud Library session has complete sets of measurements with the required calculations and graph processing steps from real assays. Instructions for running the assays are also provided. The library currently contains hundreds of protocols and continues to expand as new applications emerge.
How to use SkanIt Cloud Library?
SkanIt Cloud Library can always be accessed with the latest SkanIt version(s) when the PC is connected to Internet. The software is royalty free, so users of compatible Thermo Scientific microplate readers can freely install SkanIt Software on multiple PCs. Once SkanIt Software is installed, SkanIt Cloud Library is readily available for use. The Cloud Library icon is visible in the Home menu of the software (a), as well as in the Main menu (b), as shown below:
SkanIt microplate reader software home menu with red box around Cloud Library icon
(a)
SkanIt microplate reader software main menu with red box indicating Cloud Library icon
(b)
With the newest version of SkanIt Software, SkanIt Cloud Library has become available in a redesigned format, making it simpler to search protocols by instrument, detection technology, plate format, or other relevant search terms (if you click to enlarge the image of the search tool, you see an example in which "Invitrogen" was used as a search term). After selecting the Cloud Library, the entire list of protocols appears and the specific term of interest can be typed in the search bar. The most common search terms are already listed just below the search bar—simply click for a faster search. Additional search terms, or tags, can be easily added by clicking the [ + ] button next to the search bar to quickly customize future searches. Popular Thermo Fisher Scientific kits such as Invitrogen CyQuantInvitrogen PrestoBlue Cell Viability Reagent, or Invitrogen Quant-iT dsDNA High-Sensitivity (HS) Assay Kit can also be easily located. Some CyQuant sessions contain data with 3D spheroids.
Once the selected cloud session is opened, general technical guidance for how to perform the assay can be found in the “Notes” section. Instructions on the “Plate Layout,” “Protocol,” and “Results” sections are also included.
There are a couple of different ways to save your session, depending on your needs. If you do not need to change the protocol steps, click “Save” or “Save As” to keep the measurement data that was included in the Cloud Library session. Clicking “Save” or “Save As” makes the content of the session (except the protocol steps) editable as needed. If you need to change the protocol steps, click “Save As New” to enable that flexibility. When you click “Save As New,” the existing results data will be removed and all the content of the session will be editable as needed, including the protocol steps.
What are the benefits of using the Cloud Library?
SkanIt Cloud Library facilitates setting up assays using Thermo Scientific microplate readers and:
  • Minimizes the time needed to successfully conduct measurements, helping boost productivity.
    Learn more ways SkanIt Software helps improve productivity
  • Makes it easier to design experiments and learn how to run new protocols with technical descriptions of the assays, offering general guidance and assistance with assay performance and data processing.
    Learn more ways SkanIt Software is easy to use
  • Provides access to examples of the most frequently used microplate assays in life sciences, such as: quantification of nucleic acids, quantification of proteins, cell health (cytotoxicity, proliferation), cell signaling, reporter gene and enzyme assays, as well as ion indicators.
  • Features fully customizable protocol templates that can be customized to specified plate layouts, desired measurement, dispensing, shaking, or incubation steps, and any data processing and visualization steps needed to answer important research questions.
  • Is easily searchable via a search engine where sessions can be found quickly by using pre-defined tags or other relevant search keywords.
  • Contains hundreds of validated sessions—comprised of real assay data generated by our scientists—and the Cloud Library continues to expand as new assays and applications emerge.
Learn more about SkanIt Software
Save resources with FluoroSpot pre-screening feature of SkanIt software for microplate readers
For the first time ever, a microplate reader is able to give you an indication of the success of your FluoroSpot assay before you spend resources imaging the plate. This is a new technology offered by Thermo Scientific fluorescence microplate readers and made possible by the multipoint top-reading capability introducted with the 6th edition of SkanIt Software.
Often performed in cancer, infectious disease, and immunology research, a remarkable feature of the ELISpot assay is its sensitivity. High sensitivity is important in these fields because, for example, antigen-specific T cells are typically present at low frequencies in vivo. With ELISpot, it is possible to detect a single cytokine-producing cell from as many as one million bystander cells.
Detection of cell-secreted proteins can also be achieved using fluorescence-labeled detection reagents (including streptavidin fluorescent conjugates), and in this case the assay is known as FluoroSpot. ELISpot and FluoroSpot assays use the same protocol shown below for coating plates with capture antibodies, cell culture, and stimulation and washing steps. FluoroSpot has an advantage over ELISpot in that it is more suited for the simultaneous detection of multiple analytes, also known as multiplexing.
The ELISpot/FluoroSpot assay workflow shown graphically followed by example images of ELISpot and FluoroSpot wells
The ELISpot/FluoroSpot assay workflow.
FluoroSpot pre-screening with Thermo Scientific fluorescence microplate readers
FluoroSpot pre-screening with Thermo Scientific fluorescence microplate readers
Learn how checking your FluoroSpot plates with the Thermo Scientific exclusive pre-screening feature enables you to identify low- and high-secreting samples quickly and reduce subjectivity prior to sending for imaging.
Read the app note
Fluorescence SpectraViewer
Set up your fluorescence assays quickly and easily using the Fluorescence SpectraViewer tool.
  • The Fluorescence SpectraViewer tool—integrated in SkanIt Software—is based on the well-known and popular Fluorescence SpectraViewer on our website.
  • This dye library inside SkanIt software contains the fluorescence spectra of about 500 commonly used dyes and their excitation and emission peak wavelengths.
  • Simply choose the dye used in your assay and select the excitation and emission wavelengths for your SkanIt session, based on SpectraViewer data.
Screenshot of Fluorescence SpectraViewer tool showing the settings and excitation and emission peaks for calcium bound Fluo-4
More key features of SkanIt microplate reader software
Easy file-based user experience
  • Simply double-click SkanIt files to open them in SkanIt software—a similar experience to opening any Microsoft Office file.
  • No need for complicated import/export of SkanIt files using the latest edition of SkanIt Software.
Even faster data processing
  • Some backend software enhancements have given way to even faster performance, helping quicken your data acquisition and analysis.
PLA software interface with Stegmann Systems GmbH
  • Stegmann Systems PLA software is the standard reference for PLA calculations in the world.
  • Data is exported from SkanIt in a single click as an Excel file and imported into PLA 3.0 using Stegmann’s Microsoft Excel Data Aquisition Package. Then PLA calculations are performed using Stegmann PLA 3.0 software.
Stegmann-systems-pla-with-thermo-scientific-microplate-reader-data-from-skanit-software
Stegmann Systems PLA 3.0 software analyzing neuramidase inhibition assay data exported from SkanIt software for Thermo Scientific microplate readers.
Ordering information
All current models and some older models of Thermo Scientific microplate readers are compatible with the 6th edition of SkanIt Software. You can either download the software now or get more information by clicking the links below.
SkanIt 6 Upgrade for Fluoroskan Ascent, Fluoroskan Ascent FL, and Luminoskan Ascent models
Find out if your Fluoroskan Ascent, Fluoroskan Ascent FL, or Luminoskan Ascent can be upgraded to the 6th edition of SkanIt Software.
PC requirements
SystemPC requirements
Supported operating systems64-bit Windows 10 Professional, Education, or Enterprise edition
Hard drive14 GB disk space  
ProcessorQuad-core (or dual-core with four logical processors), 2 GHz or faster
Memory8 GB RAM
USB ports available1
MonitorFull HD with 1920 x 1080 resolution
For Research Use Only. Not for use in diagnostic procedures.