Effortless performance with every drop
Achieve unmatched speed and efficiency with Thermo Scientific Multidrop reagent dispensers. Built for high-throughput labs, Multidrop dispensers rapidly process plates with exceptional consistency—dispensing a 384-well plate in just 6 seconds and a 1536-well plate in only 15 seconds. These flexible, non-contact dispensers streamline workflows, minimize reagent waste, and keep pace with demanding screening and assay protocols. Whether you're accelerating qPCR setup or scaling drug discovery efforts, Multidrop dispensers deliver fast, reliable performance to drive productivity across your lab.
Tour the 
Multidrop Combi+
multidrop spotlight
Unleashing precision and efficiency
Precise and accurate
Provide exceptional precision and reproducibility, ensuring consistent fluid dispensing across a wide volume range. This accuracy is critical for generating reliable assay data, making them ideal for high-throughput laboratories and various applications.
Wide volume range
Offering a versatile volume range from 11 pL to 2,500 μL, accommodating a broad spectrum of applications, from low-volume qPCR to large-scale cell-based assays, ensuring flexibility and precision in every dispense.
Versatility
Dispensers are capable of handling various plate formats from 6- to 1,536-well plates, and adaptable to a wide range of applications, including drug screening, biochemical assays, and synthetic chemistry, ensuring flexibility in any laboratory setting.
Reduce reagent cost
Our dispensers minimize reagent costs with low dead volume and backflushing capabilities, significantly reducing waste and maximizing the use of expensive reagents, making them a cost-effective solution for high-throughput laboratories.
User-friendly Interface
Our dispensers’ user-friendly interface has intuitive controls and pre-programmed templates, simplifying setup and operation. This ease of use reduces training time and boosts efficiency, allowing customers to streamline their workflows effortlessly.
Automation compatible
Fully automation-friendly can seamlessly integrating with robotic systems to enhance throughput and efficiency. This compatibility makes them ideal for high-throughput laboratories seeking streamlined, automated workflows.
For Laboratory Use. It is the customer’s responsibility to ensure that the performance of the product is suitable for customers’ specific uses or applications.