Conductivity Probes
Perform your conductivity measurements with confidence
Our conductivity probes feature built-in temperature compensation; high-, standard-, and low-conductivity measuring ranges; cell constants from 0.1 to 10 cm-1, and a variety of probe materials. Our conductivity standards are NIST™ traceable and available in pouch or bottled packaging.
Popular conductivity probes products
Featured conductivity probes categories