carboys-jugs-jerricans-banner-470x200
Store and dispense solutions and media
Thermo Scientific™ Nalgene™ carboys, jugs, and jerricans are an excellent choice for the storage of reagents, solutions, media, and corrosive liquids. They're available in a variety of shapes and sizes, autoclavable, sterile, single-use, and other options—whatever your application storage need.
Popular carboys, jugs, and jerricans products
Nalgene™ Autoclavable Polypropylene Copolymer (PPCO) Lowboys with Spigot
Nalgene™ Carboy Replacement Screw Closures and Gaskets
Nalgene™ 13L HDPE Jerrican with Tethered Polypropylene Closure
Featured carboys, jugs, and jerricans categories
Store and dispense solutions and media using Nalgene Bottom Dispensing Carboys, featuring a spigot boss molded into the carboy for convenience and strength.
Use our rectangular jerricans to organize and optimize valuable workspace. Their wide stance and low center provide greater stability and better pouring control.
Store corrosive liquids in our chemical-resistant jugs which are available in HDPE, LDPE, and PP plastics and feature an offset pouring spout.
Optimize carboy use with such accessories as closures, spigots, filling/venting closures, and closures with barbed bulkhead fittings.
Store and transport media, bulk pharmaceutical ingredients, distilled water, culture media, reagents, solids, powders and liquids in our chemical-resistant carboys and tanks.
Break the glass habit
Break the glass habit
Break the habit of using glass labware and take advantage of the benefits of plastic. Plastic containers are shatterproof, lighter weight than glass, and less likely to break, which requires an expensive replacement. Make your lab a safer place and stretch your budget. Break the glass habit, switch to plastic.
WB41070