Keep your lab safe, replace your glass beakers
Safety and performance on the benchtop
Available in many different plastics and a variety of styles, including polypropylene, polymethylpentene, polyethylene, and PFA, our plastic lab beakers range in capacities from 30 mL to 4 L. Replace your glass beakers with our shatter-proof, break-, and chip-resistant lab beakers and make your lab a safer place without sacrificing accuracy.
Popular beakers products
Nalgene™ Griffin Low-Form PFA Beakers
Nalgene™ Polypropylene Griffin Low-Form Beakers
Nalgene™ PMP Griffin Low-Form Beakers
Featured beakers categories
Griffin Low-Form Beakers
Chemical resistance, heat tolerance, and spout design make Thermo Scientific™ Nalgene™ Griffin low-form beakers a perfect long-term solution for your graduated beaker choice. Meeting the ISO/DIN 7056 International Standards for Laboratory Plastic Beakers, our polypropylene and polymethylpentene beakers will provide precise measurements.
Ensure resistance against acids, bases, aliphatic alcohols, aldehydes, esters, oxidizing agents, hydrocarbons, ketones, alkalis, and hot solutions with Thermo Scientific Nalgene beakers with handles. Reliable support and dispensing control will allow you to concentrate on your research without worrying about contamination.
Beakers with Handles
Consider a convenient variety pack
Consider a convenient variety pack
Seven popular sizes in each pack—ideal for lab startup, classroom kits or lab replenishment.
Nalgene Griffin Low-Form Beaker Variety Pack
Nalgene Graduated Cylinder Variety Pack
Nalgene Interactive Guide
Break the glass habit
Resources
Certificates of Compliance
Receive a Certificate of Compliance and Certificate(s) of Irradiation (if applicable) on purchased Thermo Scientific Nalgene and Nunc products.
Start your request 
Support